Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
AVPR1B
Entrez Gene ID
553
Gene Name:
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.1372
Species:
Human
Primer Chromosome Location
Chr.1: 206116367-206115868 on Build GRCh38
View in UCSC browser
Amplicon Length
500
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00312004_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
AVPR1B
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_000707.3
NP_000698.1
4027494
c.918G>A
p.W306*
3943365
c.784C>G
p.L262V
6608972
c.713G>A
p.R238Q
2215760
c.838G>T
p.V280L
4993666
c.572_573insCGG
p.G191_F192insG
3864273
c.742G>A
p.D248N
6608970
c.784C>T
p.L262L
2215753
c.712C>T
p.R238W
6580037
c.783G>A
p.G261G
902899
c.793C>T
p.R265W
6124009
c.931C>A
p.P311T
4664797
c.714G>A
p.R238R
4664795
c.559T>C
p.W187R
4652121
c.561G>T
p.W187C
5916993
c.764T>G
p.L255*
3482372
c.636G>A
p.V212V
4551313
c.525G>A
p.R175R
1338258
c.865G>A
p.A289T
4513218
c.930C>T
p.A310A
5689867
c.886delC
p.F297fs*84
1320544
c.647C>T
p.T216M
5589772
c.919G>A
p.D307N
1235636
c.554A>G
p.D185G
1127172
c.544G>T
p.G182W
5033686
c.672T>C
p.H224H
4994034
c.568T>C
p.F190L
2215761
c.873C>T
p.I291I
3864274
c.889T>A
p.F297I
2215759
c.836C>A
p.T279K
4892943
c.934G>A
p.D312N
902900
c.859G>A
p.V287M
902898
c.570C>T
p.F190F
6045343
c.673G>T
p.E225*
4664796
c.643C>T
p.L215F
6009758
c.696C>T
p.V232V
73793
c.563C>A
p.A188E
5938573
c.592G>A
p.A198T
5845249
c.633_635delGGT
p.V212delV
1660712
c.862C>A
p.L288M
4557201
c.718G>A
p.G240R
5754327
c.781G>A
p.G261R
5705165
c.779G>A
p.R260Q
4510312
c.832C>T
p.R278*
1338257
c.835A>T
p.T279S
1237079
c.858C>T
p.I286I
69040
c.895delA
p.S299fs*82
279091
c.624C>T
p.F208F
224183
c.725G>A
p.G242E
4229980
c.552G>A
p.L184L
5368752
c.821G>A
p.R274Q
GenBank
AF030512.1
AAB84293.1
-
-
-
AF101726.1
AAD17892.1
-
-
-
D31833.1
BAA06621.1
-
-
-
ENST00000367126.0
-
-
-
EU432111.1
-
-
-
L37112.1
AAA65687.1
-
-
-
Phenotype
MIM: 600264
Literature Links:
AVPR1B
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by vasopressin
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
cellular response to hormone stimulus
positive regulation of vasoconstriction
response to peptide
vasopressin receptor activity
protein kinase C binding
peptide binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0