Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAP2K4 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001281435.1 | NP_001268364.1 | - | - | - |
NM_003010.3 | NP_003001.1 | 6965179 | c.557_558insT | p.K187fs*1 | |
975560 | c.569_591del23 | p.K190fs*47 | |||
6933912 | c.559A>T | p.K187* | |||
4700242 | c.544T>C | p.S182P | |||
6958218 | c.546_547insA | p.T183fs*4 | |||
6024069 | c.524G>A | p.W175* | |||
5702583 | c.538C>G | p.L180V | |||
5029113 | c.633+2_633+3insT | p.? | |||
7339885 | c.525G>A | p.W175* | |||
6706960 | c.617G>A | p.G206D | |||
7449684 | c.558_559insT | p.K187fs*1 | |||
6912625 | c.601C>T | p.P201S | |||
2737661 | c.607G>A | p.E203K | |||
6200095 | c.573T>A | p.Y191* | |||
1380921 | c.607G>T | p.E203* | |||
7344156 | c.587T>G | p.L196* | |||
6948415 | c.616G>T | p.G206C | |||
1380919 | c.589G>T | p.D197Y | |||
6429336 | c.632C>T | p.A211V | |||
1380920 | c.599T>C | p.I200T | |||
436035 | c.557A>G | p.D186G | |||
5226165 | c.516delT | p.D173fs*24 | |||
6945511 | c.575_578delTATA | p.Y193fs*3 | |||
436036 | c.633+1_633+2insT | p.? | |||
436034 | c.515_516insT | p.D173fs*1 | |||
6927303 | c.551C>G | p.S184W | |||
26115 | c.1_*del | p.0 | |||
1479235 | c.619A>T | p.K207* | |||
3514200 | c.602C>T | p.P201L | |||
6979303 | c.632C>A | p.A211E | |||
5802607 | c.602_604delCAG | p.P201_E202>Q | |||
3771265 | c.533T>G | p.M178R | |||
26105 | c.551C>T | p.S184L | |||
6523319 | c.592G>A | p.D198N | |||
XM_005256753.2 | XP_005256810.1 | - | - | - | |
XM_005256755.2 | XP_005256812.1 | - | - | - | |
XM_011523976.2 | XP_011522278.1 | - | - | - | |
XM_011523977.2 | XP_011522279.1 | - | - | - | |
GenBank | AK131544.1 | - | - | - | |
AK294923.1 | - | - | - | ||
AK313053.1 | - | - | - | ||
AK316100.1 | - | - | - | ||
AK316177.1 | - | - | - | ||
BC036032.1 | AAH36032.1 | - | - | - | |
BC050386.1 | - | - | - | ||
BC060764.1 | AAH60764.1 | - | - | - | |
BT019676.1 | AAV38482.1 | - | - | - | |
CR536564.1 | CAG38801.1 | - | - | - | |
ENST00000353533.0 | - | - | - | ||
L36870.1 | AAC41719.1 | - | - | - | |
U17743.1 | AAC50127.1 | - | - | - |
Literature Links: |
MAP2K4 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |