Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTNNB1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001098209.1 | NP_001091679.1 | - | - | - |
NM_001098210.1 | NP_001091680.1 | - | - | - | |
NM_001904.3 | NP_001895.1 | 6941850 | c.102_416del515 | p.? | |
4117540 | c.250G>A | p.G84R | |||
6035 | c.14_430del417 | p.A5_Q143del | |||
6034 | c.14_427del414 | p.A5_Q143>E | |||
6051 | c.64_306del243 | p.V22_T102del | |||
6033 | c.14_424del411 | p.A5_Y142>D | |||
5346975 | c.310G>T | p.D104Y | |||
6845542 | c.274C>T | p.Q92* | |||
6191447 | c.244A>T | p.I82F | |||
6048 | c.58_423del366 | p.A20_N141del | |||
5990143 | c.256T>C | p.Y86H | |||
6122 | c.103_510del408 | p.I35_K170del | |||
6063 | c.7_378del372 | p.T3_A126del | |||
6062 | c.73_420del348 | p.W25_I140del | |||
6977236 | c.283C>G | p.R95G | |||
4605020 | c.292A>G | p.M98V | |||
6059 | c.70_393del324 | p.H24_M131del | |||
6974252 | c.247G>A | p.D83N | |||
6107 | c.46_399del354 | p.P16_K133del | |||
251359 | c.7_378del1016 | p.? | |||
4117541 | c.283C>T | p.R95* | |||
5878493 | c.47_400del554 | p.? | |||
6637575 | c.253C>T | p.Q85* | |||
5993 | c.58_333del276 | p.A20_S111del | |||
6953888 | c.25_242-25del393 | p.? | |||
5985 | c.31_424>C | p.D11_Y142>H | |||
6952900 | c.14-1_242-42del388 | p.? | |||
6075 | c.88_291del204 | p.Y30_A97del | |||
7346406 | c.242-3C>T | p.? | |||
1717885 | c.47_400del354 | p.P16_K133del | |||
1717883 | c.60_455del396 | p.A21_A152del | |||
1423031 | c.269G>A | p.R90Q | |||
6126 | c.1_261del261 | p.M1_A87del | |||
6070 | c.82_402del321 | p.Q28_H134del | |||
6949869 | c.34_417del584 | p.? | |||
498637 | c.244A>G | p.I82V | |||
6096 | c.14_421del406 | p.M5_N141>D | |||
7448254 | c.242_495del254 | p.? | |||
6045 | c.44_418del375 | p.E15_I140>V | |||
6957897 | c.299C>T | p.P100L | |||
6043 | c.37_453del417 | p.A13_R151del | |||
6042 | c.29_424del396 | p.L10_N141del | |||
5995 | c.14_430del417 | p.A5_Q143del | |||
5994 | c.28_423del396 | p.L10_N141del | |||
27668 | c.14_289del276 | p.? | |||
6038 | c.1_519del519 | p.M1_V173del | |||
XM_005264886.2 | XP_005264943.1 | - | - | - | |
XM_006712983.1 | XP_006713046.1 | - | - | - | |
XM_006712984.1 | XP_006713047.1 | - | - | - | |
XM_006712985.1 | XP_006713048.1 | - | - | - | |
XM_017005738.1 | XP_016861227.1 | - | - | - | |
GenBank | AB451264.1 | - | - | - | |
AB451392.1 | - | - | - | ||
AK289932.1 | - | - | - | ||
AK294778.1 | - | - | - | ||
AK296790.1 | - | - | - | ||
AK299954.1 | - | - | - | ||
AK314776.1 | - | - | - | ||
BC058926.1 | AAH58926.1 | - | - | - | |
DA216720.1 | - | - | - | ||
ENST00000349496.0 | - | - | - | ||
X87838.1 | CAA61107.1 | - | - | - | |
Z19054.1 | CAA79497.1 | - | - | - |
Literature Links: |
CTNNB1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |