Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
UGT1A1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000463.2 | NP_000454.1 | 6816271 | c.417C>A | p.S139S |
6816273 | c.393G>A | p.L131L | |||
5396516 | c.330C>T | p.I110I | |||
1018132 | c.323G>A | p.R108H | |||
4092522 | c.341A>T | p.K114M | |||
1018133 | c.350A>C | p.K117T | |||
1018134 | c.433T>G | p.F145V | |||
1326330 | c.406C>G | p.L136V | |||
4957180 | c.356A>G | p.D119G | |||
6090968 | c.429C>A | p.S143R | |||
109793 | c.347T>A | p.I116K | |||
6816287 | c.393G>T | p.L131L | |||
233706 | c.451G>A | p.D151N | |||
1269695 | c.441C>A | p.V147V | |||
4795666 | c.325G>A | p.V109M | |||
304115 | c.449C>T | p.T150M | |||
7095703 | c.459C>T | p.F153F | |||
GenBank | AK290834.1 | - | - | - | |
AY435136.1 | AAR95637.1 | - | - | - | |
BC128414.1 | - | - | - | ||
BC128415.1 | - | - | - | ||
DQ364247.1 | - | - | - | ||
ENST00000305208.0 | - | - | - | ||
JQ686667.1 | - | - | - | ||
JQ699637.1 | - | - | - | ||
JQ699638.1 | - | - | - | ||
JQ699639.1 | - | - | - | ||
JQ699640.1 | - | - | - | ||
JQ699641.1 | - | - | - | ||
JQ699642.1 | - | - | - | ||
JQ699643.1 | - | - | - | ||
JQ699644.1 | - | - | - | ||
JQ699645.1 | - | - | - | ||
JQ699646.1 | - | - | - | ||
JQ699647.1 | - | - | - | ||
JQ699649.1 | - | - | - | ||
JQ699650.1 | - | - | - | ||
M57899.1 | AAA63195.1 | - | - | - |
UGT1A10 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_019075.2 | NP_061948.1 | - | - | - |
GenBank | AK025403.1 | - | - | - | |
AY435137.1 | AAR95638.1 | - | - | - | |
BC020971.1 | AAH20971.1 | - | - | - | |
BC053576.1 | AAH53576.1 | - | - | - | |
BC069210.1 | AAH69210.2 | - | - | - | |
ENST00000344644.0 | - | - | - | ||
U89508.1 | AAB81537.1 | - | - | - |
UGT1A3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_019093.2 | NP_061966.1 | - | - | - |
GenBank | AY435138.1 | AAR95639.1 | - | - | - |
DQ364248.1 | - | - | - | ||
ENST00000482026.0 | - | - | - |
UGT1A4 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_007120.2 | NP_009051.1 | - | - | - |
GenBank | AK313623.1 | - | - | - | |
AY435139.1 | AAR95640.1 | - | - | - | |
BC121036.1 | - | - | - | ||
BC131623.1 | - | - | - | ||
BC139784.1 | - | - | - | ||
DQ364249.1 | - | - | - | ||
ENST00000373409.0 | - | - | - | ||
M57951.1 | AAA63196.1 | - | - | - |
UGT1A5 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_019078.1 | NP_061951.1 | - | - | - |
GenBank | AY435140.1 | AAR95641.1 | - | - | - |
DQ383514.1 | - | - | - | ||
ENST00000373414.0 | - | - | - |
UGT1A6 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001072.3 | NP_001063.2 | - | - | - |
NM_205862.1 | NP_995584.1 | - | - | - | |
GenBank | AY435141.1 | AAR95642.1 | - | - | - |
BC011409.1 | - | - | - | ||
BC019861.1 | AAH19861.1 | - | - | - | |
BM688740.1 | - | - | - | ||
BM924331.1 | - | - | - | ||
DQ364250.1 | - | - | - | ||
ENST00000305139.0 | - | - | - | ||
J04093.1 | AAA61251.1 | - | - | - |
UGT1A7 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_019077.2 | NP_061950.2 | - | - | - |
GenBank | AY435142.1 | AAR95643.1 | - | - | - |
DQ383513.1 | - | - | - | ||
ENST00000373426.0 | - | - | - | ||
U89507.1 | AAB81536.1 | - | - | - |
UGT1A8 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_019076.4 | NP_061949.3 | - | - | - |
GenBank | AF030310.1 | AAB84259.1 | - | - | - |
AF462267.1 | AAL75963.1 | - | - | - | |
AF462268.1 | AAL75964.1 | - | - | - | |
AK313488.1 | - | - | - | ||
AY435143.1 | AAR95644.1 | - | - | - | |
DQ364251.1 | - | - | - | ||
ENST00000373450.0 | - | - | - |
UGT1A9 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_021027.2 | NP_066307.1 | - | - | - |
GenBank | AF056188.1 | AAC31425.1 | - | - | - |
AK313510.1 | - | - | - | ||
AY435144.1 | AAR95645.1 | - | - | - | |
BC043491.1 | - | - | - | ||
BC058844.1 | AAH58844.1 | - | - | - | |
DQ364246.1 | - | - | - | ||
ENST00000354728.0 | - | - | - | ||
S55985.1 | AAB19791.2 | - | - | - |
Literature Links: |
UGT1A1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |