Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MBD1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001204136.1 | NP_001191065.1 | - | - | - |
NM_001204137.1 | NP_001191066.1 | - | - | - | |
NM_001204138.1 | NP_001191067.1 | - | - | - | |
NM_001204139.1 | NP_001191068.1 | 5991324 | c.1446+2T>G | p.? | |
7351908 | c.1446+1G>A | p.? | |||
124344 | c.1409C>T | p.P470L | |||
282692 | c.1367T>G | p.L456R | |||
1711324 | c.1429A>T | p.T477S | |||
1659312 | c.1414C>T | p.P472S | |||
6083656 | c.1361C>G | p.T454S | |||
6026038 | c.1387G>A | p.A463T | |||
4131003 | c.1369G>T | p.V457L | |||
3378509 | c.1443G>A | p.L481L | |||
5521173 | c.1440G>T | p.L480L | |||
4467873 | c.1437C>T | p.A479A | |||
NM_001204140.1 | NP_001191069.1 | - | - | - | |
NM_001204141.1 | NP_001191070.1 | - | - | - | |
NM_001204142.1 | NP_001191071.1 | 5991326 | c.1446+2T>G | p.? | |
3378511 | c.1443G>A | p.L481L | |||
6026040 | c.1387G>A | p.A463T | |||
1711326 | c.1429A>T | p.T477S | |||
7351910 | c.1446+1G>A | p.? | |||
1659314 | c.1414C>T | p.P472S | |||
4467875 | c.1437C>T | p.A479A | |||
6083658 | c.1361C>G | p.T454S | |||
5521175 | c.1440G>T | p.L480L | |||
4131005 | c.1369G>T | p.V457L | |||
1147996 | c.1367T>G | p.L456R | |||
NM_001204143.1 | NP_001191072.1 | - | - | - | |
NM_001204151.2 | NP_001191080.1 | - | - | - | |
NM_001323942.1 | NP_001310871.1 | - | - | - | |
NM_001323947.1 | NP_001310876.1 | - | - | - | |
NM_001323949.1 | NP_001310878.1 | 6026037 | c.1462G>A | p.A488T | |
1659311 | c.1489C>T | p.P497S | |||
6083655 | c.1436C>G | p.T479S | |||
1711323 | c.1504A>T | p.T502S | |||
5521172 | c.1515G>T | p.L505L | |||
4467872 | c.1512C>T | p.A504A | |||
7351907 | c.1521+1G>A | p.? | |||
3378508 | c.1518G>A | p.L506L | |||
1147995 | c.1442T>G | p.L481R | |||
5991323 | c.1521+2T>G | p.? | |||
4131002 | c.1444G>T | p.V482L | |||
NM_001323950.1 | NP_001310879.1 | - | - | - | |
NM_001323951.1 | NP_001310880.1 | - | - | - | |
NM_001323952.1 | NP_001310881.1 | - | - | - | |
NM_001323953.1 | NP_001310882.1 | 3378510 | c.1374G>A | p.L458L | |
709409 | c.1298T>G | p.L433R | |||
6026039 | c.1318G>A | p.A440T | |||
5991325 | c.1377+2T>G | p.? | |||
4131004 | c.1300G>T | p.V434L | |||
1711325 | c.1360A>T | p.T454S | |||
7351909 | c.1377+1G>A | p.? | |||
6083657 | c.1292C>G | p.T431S | |||
1659313 | c.1345C>T | p.P449S | |||
4467874 | c.1368C>T | p.A456A | |||
5521174 | c.1371G>T | p.L457L | |||
NM_001323954.1 | NP_001310883.1 | - | - | - | |
NM_002384.2 | NP_002375.1 | - | - | - | |
NM_015844.2 | NP_056669.2 | - | - | - | |
NM_015845.3 | NP_056670.2 | - | - | - | |
NM_015846.3 | NP_056671.2 | - | - | - | |
NM_015847.3 | NP_056723.2 | - | - | - | |
XM_005258271.2 | XP_005258328.1 | - | - | - | |
XM_006722456.2 | XP_006722519.1 | - | - | - | |
XM_011525991.1 | XP_011524293.1 | - | - | - | |
XM_011525993.2 | XP_011524295.1 | - | - | - | |
XM_011525994.2 | XP_011524296.1 | - | - | - | |
XM_011525998.2 | XP_011524300.1 | - | - | - | |
XM_011525999.1 | XP_011524301.1 | - | - | - | |
XM_011526001.1 | XP_011524303.1 | - | - | - | |
XM_011526002.1 | XP_011524304.1 | - | - | - | |
XM_011526003.2 | XP_011524305.1 | - | - | - | |
XM_011526006.1 | XP_011524308.1 | - | - | - | |
XM_011526007.1 | XP_011524309.1 | - | - | - | |
XM_017025751.1 | XP_016881240.1 | - | - | - | |
XM_017025752.1 | XP_016881241.1 | - | - | - | |
XM_017025753.1 | XP_016881242.1 | - | - | - | |
XM_017025754.1 | XP_016881243.1 | - | - | - | |
XM_017025755.1 | XP_016881244.1 | - | - | - | |
XM_017025756.1 | XP_016881245.1 | - | - | - | |
XM_017025757.1 | XP_016881246.1 | - | - | - | |
XM_017025758.1 | XP_016881247.1 | - | - | - | |
XM_017025759.1 | XP_016881248.1 | - | - | - | |
XM_017025760.1 | XP_016881249.1 | - | - | - | |
XM_017025761.1 | XP_016881250.1 | - | - | - | |
XM_017025762.1 | XP_016881251.1 | - | - | - | |
XM_017025763.1 | XP_016881252.1 | - | - | - | |
XM_017025764.1 | XP_016881253.1 | - | - | - | |
XM_017025765.1 | XP_016881254.1 | - | - | - | |
XM_017025766.1 | XP_016881255.1 | - | - | - | |
XM_017025767.1 | XP_016881256.1 | - | - | - | |
XM_017025768.1 | XP_016881257.1 | - | - | - | |
XM_017025769.1 | XP_016881258.1 | - | - | - | |
XM_017025770.1 | XP_016881259.1 | - | - | - | |
XM_017025771.1 | XP_016881260.1 | - | - | - | |
XM_017025772.1 | XP_016881261.1 | - | - | - | |
XM_017025773.1 | XP_016881262.1 | - | - | - | |
XM_017025774.1 | XP_016881263.1 | - | - | - | |
XM_017025775.1 | XP_016881264.1 | - | - | - | |
XM_017025776.1 | XP_016881265.1 | - | - | - | |
XM_017025777.1 | XP_016881266.1 | - | - | - | |
GenBank | AB209765.1 | BAD93002.1 | - | - | - |
AF078830.1 | AAD51442.1 | - | - | - | |
AF078831.1 | AAD51443.1 | - | - | - | |
AF078832.1 | AAD51444.1 | - | - | - | |
AF078833.1 | AAD51445.1 | - | - | - | |
AJ564845.1 | CAD92308.1 | - | - | - | |
AK225889.1 | - | - | - | ||
AK225948.1 | - | - | - | ||
AK291519.1 | - | - | - | ||
AK295401.1 | - | - | - | ||
AK300759.1 | - | - | - | ||
AK302004.1 | - | - | - | ||
BC033242.1 | AAH33242.1 | - | - | - | |
BC047353.1 | - | - | - | ||
EF488685.1 | - | - | - | ||
ENST00000382948.0 | - | - | - | ||
Y10746.1 | CAA71735.1 | - | - | - |
Literature Links: |
MBD1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |