Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ADGRL3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001322246.1 | NP_001309175.1 | 6167849 | c.629G>T | p.G210V |
4698519 | c.504G>T | p.K168N | |||
6189046 | c.668A>T | p.E223V | |||
3302761 | c.670C>T | p.R224C | |||
4437604 | c.672C>T | p.R224R | |||
7299543 | c.522G>T | p.W174C | |||
5418085 | c.563A>G | p.K188R | |||
7375313 | c.526C>T | p.P176S | |||
1131097 | c.612C>T | p.H204H | |||
5772025 | c.527C>A | p.P176H | |||
4996451 | c.611A>T | p.H204L | |||
3373528 | c.634G>T | p.V212L | |||
7208617 | c.628G>A | p.G210R | |||
5344734 | c.671G>A | p.R224H | |||
733983 | c.587C>A | p.P196Q | |||
3409395 | c.551A>G | p.E184G | |||
6167845 | c.669G>C | p.E223D | |||
5949077 | c.549T>A | p.T183T | |||
4698523 | c.580G>T | p.G194* | |||
5975182 | c.527C>T | p.P176L | |||
351049 | c.663C>T | p.N221N | |||
7375309 | c.674C>A | p.T225N | |||
7375317 | c.641A>G | p.Y214C | |||
1056464 | c.533G>T | p.R178I | |||
5808017 | c.517C>A | p.P173T | |||
5035227 | c.658T>G | p.F220V | |||
5572404 | c.529T>C | p.Y177H | |||
733980 | c.649G>T | p.A217S | |||
733986 | c.565G>T | p.D189Y | |||
NM_001322402.1 | NP_001309331.1 | - | - | - | |
NM_015236.5 | NP_056051.2 | 4437602 | c.672C>T | p.R224R | |
6167847 | c.629G>T | p.G210V | |||
6189044 | c.668A>T | p.E223V | |||
5975180 | c.527C>T | p.P176L | |||
3373527 | c.634G>T | p.V212L | |||
4698521 | c.580G>T | p.G194* | |||
5772023 | c.527C>A | p.P176H | |||
3409394 | c.551A>G | p.E184G | |||
733979 | c.649G>T | p.A217S | |||
7375311 | c.526C>T | p.P176S | |||
1131096 | c.612C>T | p.H204H | |||
1056463 | c.533G>T | p.R178I | |||
6167843 | c.669G>C | p.E223D | |||
5949075 | c.549T>A | p.T183T | |||
4996449 | c.611A>T | p.H204L | |||
7208615 | c.628G>A | p.G210R | |||
3302759 | c.670C>T | p.R224C | |||
4698517 | c.504G>T | p.K168N | |||
5035225 | c.658T>G | p.F220V | |||
5344732 | c.671G>A | p.R224H | |||
733982 | c.587C>A | p.P196Q | |||
733985 | c.565G>T | p.D189Y | |||
5808015 | c.517C>A | p.P173T | |||
5572402 | c.529T>C | p.Y177H | |||
351048 | c.663C>T | p.N221N | |||
7299541 | c.522G>T | p.W174C | |||
7375307 | c.674C>A | p.T225N | |||
5418083 | c.563A>G | p.K188R | |||
7375315 | c.641A>G | p.Y214C | |||
XM_011531788.2 | XP_011530090.1 | - | - | - | |
XM_011531791.2 | XP_011530093.1 | - | - | - | |
XM_017007929.1 | XP_016863418.1 | - | - | - | |
XM_017007930.1 | XP_016863419.1 | - | - | - | |
XM_017007931.1 | XP_016863420.1 | - | - | - | |
XM_017007932.1 | XP_016863421.1 | - | - | - | |
XM_017007933.1 | XP_016863422.1 | 5019164 | c.816C>T | p.H272H | |
4698518 | c.708G>T | p.K236N | |||
5035226 | c.862T>G | p.F288V | |||
5949076 | c.753T>A | p.T251T | |||
5772024 | c.731C>A | p.P244H | |||
6167848 | c.833G>T | p.G278V | |||
7375308 | c.878C>A | p.T293N | |||
5975181 | c.731C>T | p.P244L | |||
7375316 | c.845A>G | p.Y282C | |||
5808016 | c.721C>A | p.P241T | |||
4342530 | c.737G>T | p.R246I | |||
4437603 | c.876C>T | p.R292R | |||
4996450 | c.815A>T | p.H272L | |||
7299542 | c.726G>T | p.W242C | |||
4698522 | c.784G>T | p.G262* | |||
7208616 | c.832G>A | p.G278R | |||
5344733 | c.875G>A | p.R292H | |||
7375312 | c.730C>T | p.P244S | |||
6167844 | c.873G>C | p.E291D | |||
5572403 | c.733T>C | p.Y245H | |||
6189045 | c.872A>T | p.E291V | |||
5418084 | c.767A>G | p.K256R | |||
4342535 | c.874C>T | p.R292C | |||
XM_017007934.1 | XP_016863423.1 | - | - | - | |
XM_017007935.1 | XP_016863424.1 | - | - | - | |
XM_017007936.1 | XP_016863425.1 | - | - | - | |
XM_017007937.1 | XP_016863426.1 | - | - | - | |
XM_017007938.1 | XP_016863427.1 | - | - | - | |
XM_017007939.1 | XP_016863428.1 | - | - | - | |
XM_017007940.1 | XP_016863429.1 | - | - | - | |
XM_017007941.1 | XP_016863430.1 | - | - | - | |
XM_017007942.1 | XP_016863431.1 | - | - | - | |
GenBank | AK309793.1 | - | - | - |
Literature Links: |
ADGRL3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |