Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
TRIM24
Entrez Gene ID
8805
Gene Name:
tripartite motif containing 24
Gene Aliases
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1
Gene Chromosome Location
Chr.7: on Build GRCh38
UniGene
Hs.490287
Species:
Human
Primer Chromosome Location
Chr.7: 138519159-138519291 on Build GRCh38
View in UCSC browser
Amplicon Length
133
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00531717_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
TRIM24
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_003852.3
NP_003843.3
4806626
c.733G>T
p.D245Y
6807980
c.656G>A
p.R219Q
6807992
c.654G>T
p.Q218H
5576173
c.706G>C
p.E236Q
326753
c.706G>A
p.E236K
334759
c.676C>G
p.H226D
1448351
c.641G>T
p.G214V
1448354
c.655C>T
p.R219*
7098854
c.676C>T
p.H226Y
5406606
c.632-1G>A
p.?
373316
c.732A>T
p.R244R
5416902
c.662T>C
p.V221A
452422
c.652C>T
p.Q218*
NM_015905.2
NP_056989.2
5416901
c.662T>C
p.V221A
326754
c.706G>A
p.E236K
7098853
c.676C>T
p.H226Y
334758
c.676C>G
p.H226D
5576172
c.706G>C
p.E236Q
4806625
c.733G>T
p.D245Y
6807979
c.656G>A
p.R219Q
1448350
c.641G>T
p.G214V
1448353
c.655C>T
p.R219*
6807991
c.654G>T
p.Q218H
452421
c.652C>T
p.Q218*
373315
c.732A>T
p.R244R
5406605
c.632-1G>A
p.?
GenBank
AF009353.1
AAB63585.1
-
-
-
AF119042.1
AAD17258.1
-
-
-
AK075306.1
-
-
-
AK302679.1
-
-
-
AK312610.1
-
-
-
BC028689.2
AAH28689.2
-
-
-
ENST00000343526.0
-
-
-
Phenotype
MIM: 603406
Literature Links:
TRIM24
PubMed Links
SNP ID
rs777662475
rs970860804
rs746543322
rs758369031
rs112052482
rs752482068
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519228-138519228 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519195-138519195 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519240-138519240 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519207-138519207 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519212-138519212 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.7: 138519204-138519204 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
protein ubiquitination
peptidyl-tyrosine phosphorylation
protein catabolic process
regulation of protein stability
regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
protein autophosphorylation
calcium ion homeostasis
regulation of vitamin D receptor signaling pathway
cellular response to estrogen stimulus
regulation of signal transduction by p53 class mediator
p53 binding
chromatin binding
transcription coactivator activity
protein tyrosine kinase activity
ubiquitin-protein transferase activity
receptor binding
protein binding
zinc ion binding
ligase activity
ligand-dependent nuclear receptor binding
estrogen response element binding
methylated histone binding
sequence-specific DNA binding
ubiquitin protein ligase activity
lysine-acetylated histone binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0