Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SCN3A | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001081676.1 | NP_001075145.1 | - | - | - |
NM_001081677.1 | NP_001075146.1 | 3311977 | c.5197G>A | p.A1733T | |
3568691 | c.5111G>A | p.S1704N | |||
3311971 | c.5267C>T | p.T1756I | |||
1664056 | c.5049C>G | p.H1683Q | |||
4934894 | c.5020C>A | p.P1674T | |||
1136367 | c.5279A>G | p.E1760G | |||
1691250 | c.5068G>A | p.G1690R | |||
1148372 | c.5258C>A | p.P1753H | |||
1224866 | c.5182G>T | p.A1728S | |||
6773028 | c.5178T>C | p.S1726S | |||
5560312 | c.5228T>C | p.M1743T | |||
6263421 | c.5247A>G | p.E1749E | |||
1153510 | c.5220C>T | p.D1740D | |||
1482105 | c.5265G>A | p.A1755A | |||
1590856 | c.5100C>A | p.F1700L | |||
5946790 | c.5247A>C | p.E1749D | |||
1325808 | c.5159T>A | p.V1720D | |||
7388871 | c.5260G>T | p.D1754Y | |||
4086131 | c.5257C>T | p.P1753S | |||
6890054 | c.5130C>T | p.F1710F | |||
1400582 | c.5157G>A | p.A1719A | |||
3568689 | c.5275A>T | p.I1759L | |||
3837204 | c.5048A>C | p.H1683P | |||
3311973 | c.5259C>T | p.P1753P | |||
6773022 | c.5264C>T | p.A1755V | |||
5476239 | c.5127C>A | p.S1709S | |||
5454925 | c.5238G>T | p.E1746D | |||
4395036 | c.5245G>A | p.E1749K | |||
4957010 | c.5271G>T | p.Q1757H | |||
6154526 | c.5024C>A | p.P1675H | |||
1265010 | c.5232C>A | p.F1744L | |||
6192461 | c.5110A>G | p.S1704G | |||
7229917 | c.5116A>G | p.I1706V | |||
1590857 | c.5118C>A | p.I1706I | |||
6563873 | c.5061A>G | p.S1687S | |||
4551260 | c.5093G>A | p.G1698E | |||
5953003 | c.5111G>C | p.S1704T | |||
3425378 | c.5156C>T | p.A1719V | |||
4592850 | c.5260G>A | p.D1754N | |||
NM_006922.3 | NP_008853.3 | 4934893 | c.5167C>A | p.P1723T | |
3568688 | c.5422A>T | p.I1808L | |||
251116 | c.5215G>A | p.G1739R | |||
335159 | c.5234C>A | p.S1745Y | |||
300670 | c.5407G>A | p.D1803N | |||
4957009 | c.5418G>T | p.Q1806H | |||
1265009 | c.5379C>A | p.F1793L | |||
6773027 | c.5325T>C | p.S1775S | |||
3837203 | c.5195A>C | p.H1732P | |||
1008515 | c.5265C>A | p.I1755I | |||
5560311 | c.5375T>C | p.M1792T | |||
4086130 | c.5404C>T | p.P1802S | |||
3425377 | c.5303C>T | p.A1768V | |||
118362 | c.5175C>T | p.D1725D | |||
6263420 | c.5394A>G | p.E1798E | |||
3311972 | c.5406C>T | p.P1802P | |||
5953002 | c.5258G>C | p.S1753T | |||
1224865 | c.5329G>T | p.A1777S | |||
7229916 | c.5263A>G | p.I1755V | |||
6773021 | c.5411C>T | p.A1804V | |||
5476238 | c.5274C>A | p.S1758S | |||
5454924 | c.5385G>T | p.E1795D | |||
3568690 | c.5258G>A | p.S1753N | |||
6154525 | c.5171C>A | p.P1724H | |||
72559 | c.5303C>G | p.A1768G | |||
7388870 | c.5407G>T | p.D1803Y | |||
6192460 | c.5257A>G | p.S1753G | |||
1008516 | c.5247C>A | p.F1749L | |||
1008514 | c.5367C>T | p.D1789D | |||
1482104 | c.5412G>A | p.A1804A | |||
1136366 | c.5426A>G | p.E1809G | |||
4551259 | c.5240G>A | p.G1747E | |||
6563872 | c.5208A>G | p.S1736S | |||
1325807 | c.5306T>A | p.V1769D | |||
3311976 | c.5344G>A | p.A1782T | |||
1664055 | c.5196C>G | p.H1732Q | |||
228875 | c.5391G>A | p.W1797* | |||
4395035 | c.5392G>A | p.E1798K | |||
3311970 | c.5414C>T | p.T1805I | |||
1400581 | c.5304G>A | p.A1768A | |||
717139 | c.5405C>A | p.P1802H | |||
5946789 | c.5394A>C | p.E1798D | |||
6890053 | c.5277C>T | p.F1759F | |||
XM_011511610.2 | XP_011509912.1 | - | - | - | |
XM_011511613.2 | XP_011509915.1 | - | - | - | |
XM_017004660.1 | XP_016860149.1 | - | - | - | |
XM_017004661.1 | XP_016860150.1 | - | - | - | |
XM_017004662.1 | XP_016860151.1 | - | - | - | |
XM_017004663.1 | XP_016860152.1 | - | - | - | |
GenBank | AB037777.1 | BAA92594.1 | - | - | - |
AF225986.1 | AAK00218.1 | - | - | - | |
AF225987.1 | AAK00219.1 | - | - | - | |
AF239921.1 | AAF44690.1 | - | - | - | |
AJ251507.1 | CAB85895.1 | - | - | - | |
AK301853.1 | - | - | - | ||
ENST00000283254.0 | - | - | - |
Literature Links: |
SCN3A PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |