Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
OR4C46
Entrez Gene ID
119749
Gene Name:
olfactory receptor family 4 subfamily C member 46
Gene Aliases
-
Gene Chromosome Location
Chr.11: on Build GRCh38
UniGene
Hs.553564
Species:
Human
Primer Chromosome Location
Chr.11: 54603790-54603545 on Build GRCh38
View in UCSC browser
Amplicon Length
246
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00660162_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
OR4C46
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001004703.1
NP_001004703.1
542804
c.341C>A
p.T114N
386387
c.424C>T
p.L142L
6069192
c.335T>A
p.L112Q
3448490
c.233A>G
p.N78S
5616530
c.267C>A
p.A89A
689126
c.449G>T
p.G150V
3448494
c.402G>A
p.M134I
3448492
c.257G>A
p.G86E
4484102
c.276C>T
p.F92F
5659876
c.407A>T
p.Q136L
1675983
c.238A>G
p.I80V
6133283
c.276C>A
p.F92L
5716525
c.430G>A
p.G144R
4033497
c.328G>T
p.G110C
6133281
c.429G>T
p.M143I
1703922
c.288G>A
p.M96I
5697730
c.286A>T
p.M96L
4033495
c.225T>C
p.N75N
4193316
c.239T>C
p.I80T
4709907
c.330C>T
p.G110G
1260255
c.338T>G
p.L113R
365625
c.435G>T
p.V145V
4193333
c.418G>T
p.A140S
4193327
c.325G>C
p.E109Q
5838728
c.418G>A
p.A140T
3716801
c.222C>A
p.V74V
5916089
c.309T>A
p.H103Q
322302
c.244C>T
p.H82Y
6734409
c.450C>A
p.G150G
4952322
c.351C>A
p.A117A
3397761
c.325G>A
p.E109K
4398070
c.233A>C
p.N78T
3769325
c.243A>C
p.T81T
3869319
c.302G>A
p.G101E
138814
c.427A>T
p.M143L
3415927
c.261G>A
p.K87K
689127
c.220G>T
p.V74F
1604660
c.345G>A
p.V115V
3448491
c.252C>T
p.L84L
689125
c.450C>T
p.G150G
6069191
c.391A>G
p.M131V
3448489
c.215C>T
p.S72F
7150355
c.213T>C
p.Y71Y
6133280
c.431G>T
p.G144V
3448493
c.267C>T
p.A89A
5701959
c.443_444TG>AT
p.M148N
4033496
c.251T>C
p.L84P
7183362
c.393G>C
p.M131I
4566674
c.440_441GG>AA
p.W147*
6133282
c.412G>T
p.V138L
7412219
c.369C>G
p.A123A
6353893
c.359A>G
p.H120R
1732460
c.419C>G
p.A140G
4033498
c.404A>C
p.N135T
5758402
c.381C>G
p.P127P
4193331
c.406C>A
p.Q136K
4922725
c.374G>T
p.C125F
4193325
c.310T>G
p.F104V
5838727
c.306A>T
p.E102D
317750
c.285C>A
p.C95*
4193334
c.431G>A
p.G144E
4953826
c.444G>T
p.M148I
1260258
c.227C>T
p.T76I
330813
c.387C>A
p.H129Q
GenBank
ENST00000328188.0
-
-
-
Phenotype
MIM: 614273
Literature Links:
OR4C46
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0