Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
GPRIN2
LOC107984026
Entrez Gene ID
9721
107984026
Gene Name:
G protein regulated inducer of neurite outgrowth 2
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Gene Aliases
GRIN2, KIAA0514
-
Gene Chromosome Location
Chr.10: on Build GRCh38
Chr.10: on Build GRCh38
UniGene
Hs.523375
Species:
Human
Primer Chromosome Location
Chr.10: 46550210-46549949 on Build GRCh38
View in UCSC browser
Amplicon Length
262
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00724114_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
GPRIN2
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_014696.3
NP_055511.2
6355149
c.584C>T
p.S195L
7420794
c.610A>T
p.T204S
4933368
c.577C>G
p.Q193E
6303995
c.722T>C
p.V241A
7351425
c.644C>A
p.P215H
6564816
c.765C>T
p.I255I
7137526
c.640G>A
p.E214K
6019120
c.673A>C
p.T225P
3931033
c.750A>C
p.L250L
3721972
c.716G>A
p.R239K
4688308
c.759A>G
p.T253T
7305610
c.718_719ins10
p.E240fs*45
3867244
c.698C>T
p.A233V
5701482
c.727_728insGA
p.A243fs*20
3867242
c.669C>T
p.T223T
5672521
c.728delC
p.A243fs*19
3438013
c.761G>A
p.G254E
1958302
c.609C>T
p.D203D
6993476
c.660G>A
p.Q220Q
5424434
c.697G>T
p.A233S
3807216
c.604G>T
p.G202W
1578430
c.720_721insATGAGGGAG
p.E240_V241insMRE
5360305
c.569G>C
p.G190A
6569207
c.655G>T
p.E219*
5633545
c.628A>G
p.S210G
4439641
c.724_725insGGGAGGTGG
p.R242_A243insEVG
6304370
c.695C>A
p.A232D
332184
c.663G>T
p.L221L
4952927
c.564G>T
p.M188I
1165883
c.721G>A
p.V241M
1958303
c.728C>A
p.A243D
3931032
c.705C>T
p.L235L
1253606
c.788C>T
p.A263V
7337316
c.769G>A
p.A257T
3721973
c.724A>G
p.R242G
4014304
c.661C>T
p.L221L
684246
c.655G>A
p.E219K
3867243
c.692C>T
p.P231L
5701451
c.712_713insATGAGGGAG
p.M238>NEGV
3438014
c.776C>T
p.P259L
5654848
c.683C>A
p.A228D
3867241
c.530C>T
p.A177V
1741083
c.721_722insATGCGGGAG
p.E240_V241insDAG
5943034
c.728_729insGG
p.G244fs*19
4594613
c.585A>G
p.S195S
3807215
c.541G>A
p.E181K
427580
c.711_712insATGAGGGAG
p.G237_M238insMRE
5341249
c.572C>A
p.A191E
3438012
c.556G>A
p.A186T
1702295
c.668C>T
p.T223I
3807217
c.642G>A
p.E214E
XM_005277666.3
XP_005277723.1
-
-
-
XM_011540388.2
XP_011538690.1
-
-
-
XM_011540395.2
XP_011538697.1
-
-
-
XM_011540400.2
XP_011538702.1
-
-
-
XM_017016974.1
XP_016872463.1
-
-
-
XM_017016975.1
XP_016872464.1
-
-
-
XM_017016976.1
XP_016872465.1
-
-
-
XM_017016977.1
XP_016872466.1
-
-
-
XM_017016978.1
XP_016872467.1
-
-
-
XM_017016979.1
XP_016872468.1
-
-
-
XM_017016980.1
XP_016872469.1
-
-
-
XM_017016981.1
XP_016872470.1
-
-
-
XM_017016982.1
XP_016872471.1
-
-
-
XM_017016983.1
XP_016872472.1
-
-
-
XM_017016984.1
XP_016872473.1
-
-
-
GenBank
AB011086.1
BAA25440.2
-
-
-
BC011672.2
AAH11672.1
-
-
-
ENST00000374317.0
-
-
-
Phenotype
MIM: 611240
LOC107984026
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
XM_017017029.1
XP_016872518.1
-
-
-
XM_017017030.1
XP_016872519.1
-
-
-
XM_017017031.1
XP_016872520.1
-
-
-
XM_017017032.1
XP_016872521.1
-
-
-
XM_017017033.1
XP_016872522.1
-
-
-
XM_017017034.1
XP_016872523.1
-
-
-
XM_017017038.1
XP_016872527.1
-
-
-
XM_017017039.1
XP_016872528.1
-
-
-
XM_017017040.1
XP_016872529.1
-
-
-
Phenotype
Literature Links:
GPRIN2
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Process(es)
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-p8qt9:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0