Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CXCR4 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001008540.1 | NP_001008540.1 | - | - | - |
NM_003467.2 | NP_003458.1 | 4303848 | c.861C>T | p.T287T | |
6948229 | c.916G>A | p.G306R | |||
7291780 | c.931A>G | p.T311A | |||
6987163 | c.995_997GAA>AAT | p.G332_K333>E* | |||
4969890 | c.946G>A | p.A316T | |||
6253527 | c.949C>A | p.L317I | |||
4926392 | c.987C>T | p.L329L | |||
6205380 | c.997A>T | p.K333* | |||
4862615 | c.842A>G | p.H281R | |||
4303847 | c.967G>T | p.G323W | |||
4085049 | c.901C>A | p.L301I | |||
5779092 | c.892A>T | p.N298Y | |||
4784693 | c.985C>A | p.L329I | |||
5394379 | c.994G>A | p.G332R | |||
4675718 | c.783C>T | p.I261I | |||
5058090 | c.854C>T | p.S285F | |||
6983887 | c.1012_1015delTCAT | p.S338fs*>14 | |||
4436176 | c.1016C>T | p.S339F | |||
6978713 | c.835A>G | p.T279A | |||
6249793 | c.975C>A | p.S325R | |||
4894087 | c.894C>T | p.N298N | |||
6153814 | c.989C>G | p.S330C | |||
4303846 | c.990C>A | p.S330S | |||
6939679 | c.788C>T | p.S263F | |||
5713580 | c.952_953insA | p.T318fs*26 | |||
4812217 | c.934T>G | p.S312A | |||
6931323 | c.802G>A | p.E268K | |||
6866744 | c.981G>A | p.K327K | |||
7173556 | c.928A>G | p.K310E | |||
4519377 | c.995G>A | p.G332E | |||
4944628 | c.1008A>G | p.G336G | |||
6212964 | c.1031_1032insTT | p.E345fs*>9 | |||
6087498 | c.1033G>C | p.E345Q | |||
4171115 | c.876_877delCT | p.F293fs*17 | |||
4848797 | c.1000C>T | p.R334* | |||
3971587 | c.1028A>G | p.E343G | |||
5628078 | c.956C>T | p.S319F | |||
4784240 | c.945C>T | p.H315H | |||
7309104 | c.990C>G | p.S330S | |||
6509778 | c.1003G>T | p.G335C | |||
GenBank | AF025375.1 | AAB81970.1 | - | - | - |
AF147204.1 | AAF00130.1 | - | - | - | |
AF348491.1 | AAK29630.1 | - | - | - | |
AK129916.1 | - | - | - | ||
AK296674.1 | - | - | - | ||
AK309641.1 | - | - | - | ||
AK309885.1 | - | - | - | ||
AK312244.1 | - | - | - | ||
AY242129.1 | AAO92296.1 | - | - | - | |
BC020968.2 | AAH20968.1 | - | - | - | |
BT006660.1 | AAP35306.1 | - | - | - | |
D10924.1 | BAA01722.1 | - | - | - | |
ENST00000409817.0 | 6948228 | c.928G>A | p.G310R | ||
4436175 | c.1028C>T | p.S343F | |||
7351382 | c.957_958insC | p.A320fs*28 | |||
6931322 | c.814G>A | p.E272K | |||
5985096 | c.1017_1018insT | p.G340fs*8 | |||
221451 | c.881T>A | p.L294Q | |||
5985092 | c.1026_1027insTAA | p.S342_S343ins* | |||
5982024 | c.1024_1024delT | p.S342fs*16 | |||
5988480 | c.1024_1025insT | p.S342fs*6 | |||
6018881 | c.1046_1047insG | p.S350fs*9 | |||
5982012 | c.1037_1038delCT | p.T346fs*3 | |||
1399595 | c.957C>T | p.H319H | |||
5981997 | c.971_972delTG | p.V324fs*23 | |||
6212963 | c.1043_1044insTT | p.E349fs*>9 | |||
6006325 | c.983C>T | p.S328F | |||
5981985 | c.1025C>A | p.S342* | |||
6019333 | c.1034_1035insT | p.T346fs*2 | |||
5991614 | c.1042_1043insT | p.S348fs*2 | |||
5713579 | c.964_965insA | p.T322fs*26 | |||
6939678 | c.800C>T | p.S267F | |||
7347772 | c.1029_1030insTT | p.V344fs*15 | |||
4085048 | c.913C>A | p.L305I | |||
5985095 | c.1024_1027delTCAT | p.S342fs*15 | |||
7173555 | c.940A>G | p.K314E | |||
6866743 | c.993G>A | p.K331K | |||
204665 | c.1012C>T | p.R338* | |||
5982022 | c.1033_1033delT | p.S345fs*13 | |||
4926391 | c.999C>T | p.L333L | |||
5988479 | c.1018G>T | p.G340* | |||
6018880 | c.1006G>T | p.G336* | |||
5982011 | c.1028_1029delCT | p.S343fs*4 | |||
1399594 | c.997C>A | p.L333I | |||
6018869 | c.990_991insT | p.K331fs*1 | |||
5981995 | c.964_968delACCTC | p.T322fs*24 | |||
4675717 | c.795C>T | p.I265I | |||
6196573 | c.988_989insC | p.L330fs*18 | |||
6005793 | c.1043_1043delC | p.S348fs*10 | |||
5981983 | c.1009A>T | p.K337* | |||
6019332 | c.994_995delAT | p.I332fs*15 | |||
5991609 | c.1028_1029insC | p.V344fs*4 | |||
5779091 | c.904A>T | p.N302Y | |||
5991603 | c.1005_1006insA | p.G336fs*12 | |||
5628077 | c.968C>T | p.S323F | |||
418563 | c.946T>G | p.S316A | |||
3116600 | c.979G>T | p.G327W | |||
7309103 | c.1002C>G | p.S334S | |||
5985094 | c.963_964insTC | p.T322fs*4 | |||
716722 | c.854A>G | p.H285R | |||
5982026 | c.1039_1040ins11 | p.E347fs*15 | |||
1582480 | c.1039G>A | p.E347K | |||
5982015 | c.1046_1047delAG | p.E349fs*9 | |||
6987162 | c.1007_1009GAA>AAT | p.G336_K337>E* | |||
4894086 | c.906C>T | p.N302N | |||
5988475 | c.975_976insC | p.R326fs*22 | |||
6018879 | c.996_997insT | p.L333fs*15 | |||
5982008 | c.1026_1042del17 | p.S343fs*1 | |||
6978712 | c.847A>G | p.T283A | |||
6253526 | c.961C>A | p.L321I | |||
6018868 | c.981_982insG | p.S328fs*20 | |||
5981991 | c.944_945insT | p.S316fs*32 | |||
6153813 | c.1001C>G | p.S334C | |||
354870 | c.841G>C | p.E281Q | |||
4519376 | c.1007G>A | p.G336E | |||
6019331 | c.989_990insC | p.K331fs*17 | |||
1006910 | c.873C>T | p.T291T | |||
5991604 | c.1028_1029insT | p.V344fs*4 | |||
4171114 | c.888_889delCT | p.F297fs*17 | |||
3116599 | c.1002C>A | p.S334S | |||
5985097 | c.1021_1024delCATT | p.S342fs*15 | |||
7291779 | c.943A>G | p.T315A | |||
5394378 | c.1006G>A | p.G336R | |||
3971586 | c.1040A>G | p.E347G | |||
5985093 | c.1033_1034insT | p.S345fs*3 | |||
5058089 | c.866C>T | p.S289F | |||
5982025 | c.989T>C | p.L330P | |||
4944627 | c.1020A>G | p.G340G | |||
6018882 | c.1033_1034delTC | p.S345fs*2 | |||
5982014 | c.1043_1052CTGAGTCTTC>GT | p.S348fs*8 | |||
6509777 | c.1015G>T | p.G339C | |||
6018878 | c.943_944insT | p.T315fs*33 | |||
5982007 | c.1026_1029delATCT | p.S343fs*14 | |||
6249792 | c.987C>A | p.S329R | |||
1249498 | c.958G>A | p.A320T | |||
6018867 | c.966_967insC | p.S323fs*25 | |||
5981986 | c.1025C>G | p.S342* | |||
350375 | c.1045G>C | p.E349Q | |||
KU245646.1 | - | - | - | ||
KU245647.1 | - | - | - | ||
L01639.1 | AAA16594.1 | - | - | - | |
L06797.1 | AAA03209.1 | - | - | - | |
M99293.1 | AAA16617.1 | - | - | - | |
X71635.1 | CAA50641.1 | - | - | - |
Literature Links: |
CXCR4 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |