Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RIMS2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001100117.2 | NP_001093587.1 | - | - | - |
NM_001282881.1 | NP_001269810.1 | - | - | - | |
NM_014677.4 | NP_055492.3 | - | - | - | |
XM_005251106.3 | XP_005251163.1 | - | - | - | |
XM_005251107.3 | XP_005251164.1 | - | - | - | |
XM_011517395.2 | XP_011515697.1 | - | - | - | |
XM_011517398.2 | XP_011515700.1 | - | - | - | |
XM_017014006.1 | XP_016869495.1 | - | - | - | |
XM_017014007.1 | XP_016869496.1 | - | - | - | |
XM_017014008.1 | XP_016869497.1 | - | - | - | |
XM_017014009.1 | XP_016869498.1 | - | - | - | |
XM_017014010.1 | XP_016869499.1 | 5278712 | c.260T>C | p.V87A | |
1453746 | c.126C>T | p.T42T | |||
5507152 | c.213T>C | p.Y71Y | |||
220430 | c.127G>A | p.A43T | |||
7387545 | c.306G>T | p.R102S | |||
241421 | c.93A>T | p.R31S | |||
1223777 | c.137G>T | p.R46I | |||
131396 | c.260T>A | p.V87E | |||
3643516 | c.217G>A | p.D73N | |||
5496538 | c.102T>C | p.Y34Y | |||
6549595 | c.214A>T | p.R72W | |||
3270219 | c.247A>G | p.K83E | |||
1264253 | c.232A>C | p.S78R | |||
5592885 | c.164G>A | p.R55Q | |||
6766644 | c.163C>T | p.R55* | |||
1488865 | c.281G>A | p.S94N | |||
748960 | c.91A>G | p.R31G | |||
3884341 | c.157G>C | p.D53H | |||
6111607 | c.294C>G | p.Y98* | |||
5345458 | c.295G>A | p.E99K | |||
1453751 | c.294C>T | p.Y98Y | |||
6256905 | c.277G>T | p.E93* | |||
6053473 | c.156T>G | p.A52A | |||
216082 | c.104C>T | p.S35L | |||
6516447 | c.172C>A | p.H58N | |||
382553 | c.120G>A | p.S40S | |||
5440973 | c.181C>T | p.Q61* | |||
346027 | c.164G>T | p.R55L | |||
1094525 | c.145T>C | p.S49P | |||
4628696 | c.237T>C | p.H79H | |||
4563546 | c.313G>A | p.E105K | |||
5738411 | c.134C>A | p.P45H | |||
5538255 | c.277G>A | p.E93K | |||
6200793 | c.153T>A | p.Y51* | |||
3982332 | c.167C>T | p.S56F | |||
1623456 | c.299G>C | p.R100T | |||
1094529 | c.239G>A | p.R80K | |||
1223763 | c.119C>T | p.S40L | |||
4759710 | c.308G>C | p.R103T | |||
4428878 | c.270G>A | p.E90E | |||
6549590 | c.170A>T | p.Q57L | |||
3643509 | c.97G>A | p.E33K | |||
5894887 | c.143C>T | p.P48L | |||
3778804 | c.146C>G | p.S49* | |||
XM_017014011.1 | XP_016869500.1 | - | - | - | |
XM_017014012.1 | XP_016869501.1 | - | - | - | |
XM_017014013.1 | XP_016869502.1 | - | - | - | |
XM_017014014.1 | XP_016869503.1 | - | - | - | |
XM_017014015.1 | XP_016869504.1 | - | - | - | |
XM_017014016.1 | XP_016869505.1 | - | - | - | |
XM_017014017.1 | XP_016869506.1 | - | - | - | |
XM_017014018.1 | XP_016869507.1 | - | - | - | |
XM_017014019.1 | XP_016869508.1 | - | - | - | |
XM_017014020.1 | XP_016869509.1 | - | - | - | |
XM_017014021.1 | XP_016869510.1 | - | - | - | |
XM_017014022.1 | XP_016869511.1 | - | - | - | |
XM_017014023.1 | XP_016869512.1 | - | - | - | |
XM_017014024.1 | XP_016869513.1 | - | - | - | |
XM_017014025.1 | XP_016869514.1 | - | - | - | |
XM_017014026.1 | XP_016869515.1 | - | - | - | |
XM_017014027.1 | XP_016869516.1 | - | - | - | |
XM_017014028.1 | XP_016869517.1 | - | - | - | |
XM_017014029.1 | XP_016869518.1 | - | - | - | |
XM_017014030.1 | XP_016869519.1 | - | - | - | |
XM_017014031.1 | XP_016869520.1 | - | - | - | |
XM_017014032.1 | XP_016869521.1 | - | - | - | |
XM_017014033.1 | XP_016869522.1 | - | - | - | |
XM_017014034.1 | XP_016869523.1 | - | - | - | |
XM_017014035.1 | XP_016869524.1 | - | - | - | |
XM_017014036.1 | XP_016869525.1 | - | - | - | |
XM_017014037.1 | XP_016869526.1 | - | - | - | |
XM_017014038.1 | XP_016869527.1 | - | - | - | |
XM_017014039.1 | XP_016869528.1 | - | - | - | |
XM_017014040.1 | XP_016869529.1 | - | - | - | |
XM_017014041.1 | XP_016869530.1 | - | - | - | |
XM_017014042.1 | XP_016869531.1 | - | - | - | |
XM_017014043.1 | XP_016869532.1 | - | - | - | |
XM_017014044.1 | XP_016869533.1 | - | - | - | |
XM_017014045.1 | XP_016869534.1 | - | - | - | |
XM_017014046.1 | XP_016869535.1 | - | - | - | |
GenBank | AB018294.1 | - | - | - | |
AK126939.1 | - | - | - | ||
AK307790.1 | - | - | - | ||
AK308831.1 | - | - | - | ||
BC018057.1 | AAH18057.1 | - | - | - | |
BC043144.2 | AAH43144.1 | - | - | - | |
BP230765.1 | - | - | - | ||
CR749374.1 | CAH18227.1 | - | - | - | |
ENST00000507740.0 | 4759712 | c.398G>C | p.R133T | ||
216080 | c.194C>T | p.S65L | |||
3643518 | c.307G>A | p.D103N | |||
1623458 | c.389G>C | p.R130T | |||
4428880 | c.360G>A | p.E120E | |||
5278714 | c.350T>C | p.V117A | |||
346029 | c.254G>T | p.R85L | |||
1223779 | c.227G>T | p.R76I | |||
6549592 | c.260A>T | p.Q87L | |||
382555 | c.210G>A | p.S70S | |||
5592887 | c.254G>A | p.R85Q | |||
3778806 | c.236C>G | p.S79* | |||
241419 | c.183A>T | p.R61S | |||
5738413 | c.224C>A | p.P75H | |||
5345460 | c.385G>A | p.E129K | |||
3884343 | c.247G>C | p.D83H | |||
6256907 | c.367G>T | p.E123* | |||
5496540 | c.192T>C | p.Y64Y | |||
1094527 | c.235T>C | p.S79P | |||
5507154 | c.303T>C | p.Y101Y | |||
3982334 | c.257C>T | p.S86F | |||
5440975 | c.271C>T | p.Q91* | |||
6549597 | c.304A>T | p.R102W | |||
6516449 | c.262C>A | p.H88N | |||
1453748 | c.216C>T | p.T72T | |||
6766646 | c.253C>T | p.R85* | |||
131395 | c.350T>A | p.V117E | |||
6053475 | c.246T>G | p.A82A | |||
1264255 | c.322A>C | p.S108R | |||
1094531 | c.329G>A | p.R110K | |||
1223765 | c.209C>T | p.S70L | |||
6111609 | c.384C>G | p.Y128* | |||
6200795 | c.243T>A | p.Y81* | |||
3270221 | c.337A>G | p.K113E | |||
220428 | c.217G>A | p.A73T | |||
1453753 | c.384C>T | p.Y128Y | |||
5894889 | c.233C>T | p.P78L | |||
4628698 | c.327T>C | p.H109H | |||
3643511 | c.187G>A | p.E63K | |||
7387544 | c.396G>T | p.R132S | |||
5538257 | c.367G>A | p.E123K | |||
4563548 | c.403G>A | p.E135K | |||
1488867 | c.371G>A | p.S124N | |||
748962 | c.181A>G | p.R61G |
Literature Links: |
RIMS2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |