4 Micron Small Particle Size IC Columns
Optimize speed and resolution in ion chromatography
Now you can optimize the speed and resolution of your run using small particle Thermo Scientific Dionex IonPac 4 micron (µm) IC columns. Column efficiency doubles when particle size is reduced from 8µm to 4µm. Available in various lengths, Dionex IonPac 4µm IC columns allow you to optimize faster runs with better chromatographic resolution. Dionex HPIC high pressure ion chromatography systems were designed to operate comfortably at the higher backpressures 4μm IC columns create.
Small particle IC columns
Dionex IC Columns for High Pressure Ion Chromatography
IC Column Application Format
Dionex IonPac AS28-4µm Fast, high-resolution of trace concentrations of low molecular weight organic acids (glycolate, acetate) and inorganic anions Standard, microbore, and capillary
Dionex IonPac AS23-4µm High-resolution separation of oxyhalides and inorganic anions in diverse matrices Standard and microbore
Dionex IonPac AS11-HC-4µm High-resolution separation of organic acids and inorganic anions in complex sample matrices Standard, microbore, and capillary
Dionex IonPac AS18-4µm Fast, high-resolution separation of inorganic anions in diverse matrices Standard, microbore, and capillary
Dionex IonPac CS19-4µm Fast, high-resolution separation of cations and small polar amines in diverse matrices Standard, microbore, and capillary
Dionex IonPac CS16-4µm High-resolution separation of short chain amines and disparate ratios of sodium and ammonium Standard, microbore, and capillary
Dionex IonPac CS16-Fast-4µm Fast, high-resolution separation of short chain amines and disparate ratios of sodium and ammonium Standard, microbore, and capillary
Dionex IonPac CS12A-5µm High-efficiency and fast analysis of mono- and divalent cations Standard, microbore, and capillary
Dionex CarboPac SA10-4µm IC Columns Direct quantification of nonderivatized carbohydrates in biofuels and food samples with minimal sample preparation using pulsed amperometric detection Standard and microbore
Dionex CarboPac SA10-Fast-4µm IC Columns Fast, direct quantification of nonderivatized carbohydrates in biofuels and food samples with minimal sample preparation using pulsed amperometric detection Standard and microbore
Dionex IonPac AS22-Fast-4µm Fast, high-resolution separation of common inorganic anions Standard, microbore, and capillary
Dionex IonPac AS16-4µm High-resolution separation of polarizable anions in diverse matrices Standard, microbore, and capillary
Dionex IonPac AS19-4µm High-resolution separation of trace oxyhalides and inorganic anions in diverse matrices Standard, microbore, and capillary
Dionex CarboPac PA210-Fast-4µm Fast, high-resolution separation of mono-, di-, tri-, tetra-, and pentasaccharides in food and beverage matrices Standard and microbore
Dionex CarboPac PA20-Fast-4µm Fast, high-resolution separation of mono- and disaccharides in diverse matrices Standard and microbore
High Pressure Ion Chromatography (HPIC) systems
Learn more about Dionex HPIC systems that operate comfortably at the higher backpressures 4µm IC columns create.
Featured video
Dionex IonPac 4µm Columns
Take just a few minutes to learn about 4µm small-particle IC columns and their application.
Advancing Ion Chromatography with High Pressure eBook
HPLC users have seen significant advantages by using high pressure systems and small particle size columns. That high pressure has now come to ion chromatography delivering more powerful detection.
  • Why high-pressure IC?
  • What's the advantage of small column particle size
  • Puts the trend into context
Resources