CRISPR gRNA
High quality gRNAs for any CRISPR application
When using the CRISPR-Cas9 system to knockout gene expression or knock-in a specific mutation, the design, production, and delivery of high quality gRNAs are critical to achieving a successful result. The scientists at Thermo Fisher Scientific  have developed multiple CRISPR gRNA solutions to help you realize your goals and develop high impact models to move your research forward. Whether you need transfection-ready gRNAs for use with Invitrogen TrueCut Cas9 Protein v2 or you need to harness lentivirus to deliver your editing tools to hard to transfect or primary cells, we have you covered. Use our selection guide below to find the right gRNAs for your experiment.
Available formats
TrueGuide Synthetic gRNA
LentiArray Lentiviral gRNA
IVT gRNA Custom Service
Overview Choose from our catalog of predesigned gRNAs or upload your sequence to our Invitrogen TrueGuide gRNA Ordering Tool Choose your gRNA design from our catalog of gRNAs packaged as ready to use lentivirus. Utilize our in-house expertise and have our GeneArt custom services team design and synthesize ready to transfect gRNAs for you or design them yourselves using our design tool.
Format Synthetic gRNA oligo Lentivirus lentiviral gRNA In vitro (IVT) gRNA
Application Knockout or knock-in Knockout or library screening Knockout or knock-in
Species All species Human All species
Delivery method Lipid-mediated transfection or electroporation Lentiviral delivery Lipid-mediated transfection or electroporation
Recommended Cas9 Format Invitrogen TrueCut Cas9 Protein v2 Invitrogen LentiArray Cas9 Lentivirus Invitrogen TrueCut Cas9 Protein v2
Controls Positive and negative controls available Positive and negative controls available Available upon request
  Order now Order now Order now
CRISPR Controls
High-quality controls play an integral role in the successful development and performance of high-throughput screens. Don't forget to include controls to help you optimize delivery conditions, maximize editing efficiency, and establish hit selection criteria. Learn more
Can’t find what you are looking for? Contact us at CRISPR@thermofisher.com
Resources
  • Learning Center
    Access genome editing application resources for more success as you plan and execute your experiments.
  • CRISPR Validated Protocols
    Getting started with CRISPR? Need proven CRISPR validated protocols? Check out our collection of step-by-step validated protocols.
  • FAQs
    Find answers to everyday problems, we have consolidated a list of most commonly asked questions.
  • Webinar Series
    Master the art of CRISPR editing by tuning into our on-demand 3 part webinar series.
  • CRISPR Hands-on Workshop
    Our experienced team has designed a comprehensive four-day workshop to get you up to speed on CRISPR.
Support