Absolutely attainable chip-based digital PCR
Now, you can go beyond the limits of real-time PCR with the new Applied Biosystems QuantStudio 3D Digital PCR System. A simple and affordable platform, the QuantStudio 3D Digital PCR System produces absolute quantification data with the precision and sensitivity you need for your more demanding applications. The QuantStudio 3D Digital PCR System uses a sealed chip technology, providing a streamlined, reliable, and robust method for performing digital PCR. Just load and go.
Request a quote  Technical support
A new generation of PCR is here
The QuantStudio 3D Digital PCR System is:
  • Simple—load and go, turn-key chip-based workflow
  • Affordable—less than 50% of the price of competing platforms
  • Absolute—concentration measurements in copies/μL without a standard curve
Watch a workflow video to see the simplicity of QuantStudio 3D Digital PCR System.
Simple workflow limits hands-on steps and sample manipulation
The QuantStudio 3D Digital PCR Chip enables a simple and streamlined workflow. Unlike other methods that require multiple transfer steps and upfront sample digestion, the QuantStudio 3D Digital PCR Chip is simply loaded and sealed—ready for PCR and analysis. With the ability to thermal cycle up to 24 chips at one time and taking less than 30 seconds to read each chip, the QuantStudio 3D Digital PCR System enables a simple, streamlined workflow to perform digital PCR.
QuantStudio 3D workflow
Watch a workflow video to see the simplicity of QuantStudio 3D Digital PCR System.
Lab Genomics, Inc of Korea uses QuantStudio 3D Digital PCR system to ID low frequency mutations in FFPE samples.
Catholic University of Korea uses the dPCR to detect low frequency mutations implicated in prostate and lung cancer. 
Affordable solution to enable digital PCR in any lab
Because the QuantStudio 3D Digital PCR System is not based on expensive droplet flow technology, it is able to be priced significantly lower than other systems on the market. Compared to the closest digital PCR competitor, it would take at least 10 years of average use* to equal the savings achieved with the QuantStudio 3D Digital PCR System. In addition, as the provider of over 8 million Applied Biosystems™ TaqMan® Assays, a staple of digital PCR, we can maximize your savings for the assays you need.
Learn about our specially-priced comprehensive package, designed to help you get started with digital PCR in your lab right away.
Absolute quantification at your fingertips
Absolute quantification by digital PCR is made possible by thousands of individual reactions running in parallel. Manufactured from a silicon substrate and etched with 20,000 consistently sized nanoscale reaction wells, the Applied Biosystems™ QuantStudio™ 3D Digital PCR Chip enables more controlled sample loading and less reaction dropout than droplet-based approaches, ensuring higher precision for absolute quantification.
Digital PCR extends the performance of existing TaqMan Assays, enabling applications that benefit from higher sensitivity, precision, or absolute quantification. Such applications include rare mutation detection (otherwise known as looking for a needle in a haystack such as in cancer samples), virus and bacteria quantification and detection (getting an absolute count of pathogens detected), genetically-modified organism detection (detecting a GMO seed where in places it shouldn’t be), next-generation sequencing workflow optimization (accurately quantifying NGS libraries and validating sequencing results), and reference standard quantification (developing a reference for your real‐time experiment when it doesn’t exist).
To learn more, watch this webinar “Expanding the PCR toolbox with Digital PCR – A review of absolute quantification and mutation detection applications on the QuantStudio 3D Digital PCR System”
Chose from the applications below to learn more:
Manuals & Protocols
User Bulletin: Supported Protocol: Getting Started with the QuantStudio 3D Digital PCR System
User Guide: QuantStudio 3D Digital PCR System User Guide
User Guide: QuantStudio 3D AnalysisSuite Software User Guide
Quick Reference: QuantStudio 3D Digital PCR System
Product Literature
QuantStudio 3D Digital PCR System Product Bulletin
QuantStudio 3D Digital PCR System Experiment Design Guide
Release Notes: QuantStudio 3D Digital PCR Software v3.0
Application Literature
Copy Number Variation (CNV) Using Digital PCR
Your Innovative Research: "Copy number variation in breast cancer translational research; QuantStudio 3D Digital PCR System as a cost-effective and sensitive alternative for HER2 gene amplification assessment"
Copy Number Variation Analysis Using the QuantStudio 3D Digital PCR System
Next Generation Sequencing (NGS) Library Quantification
Poster: Precise Quantification of Next Generation Sequencing Ion Torrent Libraries with the QuantStudio 3D Digital PCR Platform
Precise Quantification of Ion Torrent Libraries on the QuantStudio 3D Digital PCR System
Absolute Quantification
Application Note: DNA Quantitation of Meat Species by Digital PCR
Differential Gene Expression Using Digital PCR
Poster: Precise and Accurate Determination of MicroRNA Precursors by Digital PCR
Rare Mutation Analysis
Poster: Multiplex TaqMan Assays for Rare Mutation Analysis Using Digital PCR
Fusion Transcript Detection
Fusion Transcript Detection with Digital PCR
Simple workflow limits hands-on steps and sample manipulation
The QuantStudio 3D Digital PCR Chip enables a simple and streamlined workflow. Unlike other methods that require multiple transfer steps and upfront sample digestion, the QuantStudio 3D Digital PCR Chip is simply loaded and sealed—ready for PCR and analysis. With the ability to thermal cycle up to 24 chips at one time and taking less than 30 seconds to read each chip, the QuantStudio 3D Digital PCR System enables a simple, streamlined workflow to perform digital PCR.
QuantStudio 3D workflow
Watch a workflow video to see the simplicity of QuantStudio 3D Digital PCR System.
Lab Genomics, Inc of Korea uses QuantStudio 3D Digital PCR system to ID low frequency mutations in FFPE samples.
Catholic University of Korea uses the dPCR to detect low frequency mutations implicated in prostate and lung cancer. 
Affordable solution to enable digital PCR in any lab
Because the QuantStudio 3D Digital PCR System is not based on expensive droplet flow technology, it is able to be priced significantly lower than other systems on the market. Compared to the closest digital PCR competitor, it would take at least 10 years of average use* to equal the savings achieved with the QuantStudio 3D Digital PCR System. In addition, as the provider of over 8 million Applied Biosystems™ TaqMan® Assays, a staple of digital PCR, we can maximize your savings for the assays you need.
Learn about our specially-priced comprehensive package, designed to help you get started with digital PCR in your lab right away.
Absolute quantification at your fingertips
Absolute quantification by digital PCR is made possible by thousands of individual reactions running in parallel. Manufactured from a silicon substrate and etched with 20,000 consistently sized nanoscale reaction wells, the Applied Biosystems™ QuantStudio™ 3D Digital PCR Chip enables more controlled sample loading and less reaction dropout than droplet-based approaches, ensuring higher precision for absolute quantification.
Digital PCR extends the performance of existing TaqMan Assays, enabling applications that benefit from higher sensitivity, precision, or absolute quantification. Such applications include rare mutation detection (otherwise known as looking for a needle in a haystack such as in cancer samples), virus and bacteria quantification and detection (getting an absolute count of pathogens detected), genetically-modified organism detection (detecting a GMO seed where in places it shouldn’t be), next-generation sequencing workflow optimization (accurately quantifying NGS libraries and validating sequencing results), and reference standard quantification (developing a reference for your real‐time experiment when it doesn’t exist).
To learn more, watch this webinar “Expanding the PCR toolbox with Digital PCR – A review of absolute quantification and mutation detection applications on the QuantStudio 3D Digital PCR System”
Chose from the applications below to learn more:
Manuals & Protocols
User Bulletin: Supported Protocol: Getting Started with the QuantStudio 3D Digital PCR System
User Guide: QuantStudio 3D Digital PCR System User Guide
User Guide: QuantStudio 3D AnalysisSuite Software User Guide
Quick Reference: QuantStudio 3D Digital PCR System
Product Literature
QuantStudio 3D Digital PCR System Product Bulletin
QuantStudio 3D Digital PCR System Experiment Design Guide
Release Notes: QuantStudio 3D Digital PCR Software v3.0
Application Literature
Copy Number Variation (CNV) Using Digital PCR
Your Innovative Research: "Copy number variation in breast cancer translational research; QuantStudio 3D Digital PCR System as a cost-effective and sensitive alternative for HER2 gene amplification assessment"
Copy Number Variation Analysis Using the QuantStudio 3D Digital PCR System
Next Generation Sequencing (NGS) Library Quantification
Poster: Precise Quantification of Next Generation Sequencing Ion Torrent Libraries with the QuantStudio 3D Digital PCR Platform
Precise Quantification of Ion Torrent Libraries on the QuantStudio 3D Digital PCR System
Absolute Quantification
Application Note: DNA Quantitation of Meat Species by Digital PCR
Differential Gene Expression Using Digital PCR
Poster: Precise and Accurate Determination of MicroRNA Precursors by Digital PCR
Rare Mutation Analysis
Poster: Multiplex TaqMan Assays for Rare Mutation Analysis Using Digital PCR
Fusion Transcript Detection
Fusion Transcript Detection with Digital PCR
Digital PCR Augmented Reality App
Watch the instrument come alive in 3D right on your lab bench. Take a virtual tour of the QuantStudio 3D Digital PCR System to learn about the instrument's innovative features and sealed, chip-based workflow.
Get the app
Keep informed
Watch our digital PCR webinar series to learn more about digital PCR and its applications.