Tradeshow
Sep 25, 2017 - Sep 27, 2017
Las Vegas Convention Center  | 3150 Paradise Rd, Las Vegas, NV 89109
Going to Pack Expo in September?
Visit us at Booth C-2200 and discover the latest product inspection innovations in checkweighing, metal detection, and X-ray inspection. Protect your brand and ensure the safety and quality of your packaged and unpackaged products.
HDI-Pack-Expo-2017-270x195
Featured Product: Our new Thermo Scientific™ Sentinel™ 5000 Multiscan Metal Detector
Be among the first to see our new Multiscan technology for metal detectors. It's in our Sentinel metal detector launching at Pack Expo. Our Multiscan technology is designed to overcome the limitations of fixed 1-2 frequency metal detectors. Scan up to five user-selectable frequencies at a time for unmatched sensitivity and the highest probability of detection. Stop by for a demo.
Sentinel
HDI-Pack-Expo-Presenter-Bob-Ries-270x195
Join us at the Pack Expo Innovation Stage, C-1049
Wednesday, September 27 at 10 a.m.
"How Multiscan Technology Will Revolutionize Metal Detection and Food Safety"
Presented by Bob Ries, lead product manager, Metal Detection and X-ray Inspection, Thermo Fisher Scientific
Metal detection has been limited to one or two frequencies due to technical constraints in the past with the result being that small, real-world contaminants can escape detection. A totally new approach to this old problem will be presented that utilizes a true spectrum of frequencies along with new signal processing, thus reducing the probability of an escape to near zero. This technology will be compared to currently deployed systems highlighting the benefits of upgrading and enhancing food safety.
Catch issues before they cross the line with our product inspection solutions
Discover our latest product inspection innovations in checkweighing, metal detection, and X-ray inspection to help protect your brand and ensure the safety and quality of your packaged and unpackaged products.
Find out how our online measurement and control systems can help to optimize product quality and guarantee the performance of your materials, while minimizing raw material waste. Our product inspection experience spans hundreds of applications and our solutions are proven in over 30,000 installations.
Do you need to improve efficiency and profitability for dry applications?
The Thermo Scientific™ Versa Flex Checkweigher enables you to improve accuracy and reliability while decreasing product giveaway. These models featuring completely new checkweigher HMI provide accurate weight control for dry applications – whether packaged food, pharmaceutical or personal care products. Weigh light to heavy-weight packages.
Learn more ›
Do you want to transition from metal detection to X-ray Inspection?
Designed for a wide variety of food applications, the easy-to-use Thermo Scientific™ NextGuard™ X-ray Detection System detects a wide range of foreign objects and offers enhanced capabilities to inspect packaged products for missing pieces or components, under and over-fills, and other quality problems with easily customizable vision software.
Learn more ›
Are you looking for a metal detector providing high sensitivity and long-term stability?
Eliminate downtime, service costs, and repairs with the Thermo Scientific™ APEX HD Metal Detector. It withstands harsh washdown, mechanical vibration, and sanitation with caustic chemicals. It keeps throughput high with no false reject delays. Low cost of ownership; 3-year warranty.
Learn more ›
Learn more about our thickness and basis weight measurement solutions
Do you need accurate thickness and basis weight data for your extruded films, coatings and packaging to improve product quality and reduce raw material waste?
Extend the shelf life of food products and protect medical instruments from contaminants. The Thermo Scientific™ 21Plus! Measurement and Control System is the perfect solution for biax and cast film extrusion, extrusion and tandem extrusion coating, flexible and aseptic packaging and technical films. In conjunction with the Thermo Scientific™ PROSIS™ Infrared Process Analysis Thickness Sensor, which can measure the individual layers of a multilayer product, it is ideal for producing barrier layer films for the food, medical, and specialty film markets.
Learn more ›