Tradeshow
May 29, 2018 - Jun 01, 2018
FIERA MILANO Fairgrounds | Strada Statale 33 del Sempione, 28 - 20017 Rho (Milan), Italy
To meet ever changing end-use requirements, polymer manufacturers need to develop new materials and deliver high quality products. At any stage of the value chain, you can rely on our comprehensive solutions to speed the development, production, quality testing and analysis of polymers and plastics.
Visit us at Plast 2018 in Hall 11, Booth C106 to discover how our innovations can help solve your plastic and polymer challenges. Or stay tuned with our electronic sneak peek in the next weeks that comes to your inbox.
Discover the many solutions we offer:
Need to enhance chemical identification to gain deeper insight into your sample?
Reveal more data for enhanced chemical identification with the Thermo Scientific Nicolet iS50 FTIR Spectrometer. Build your ideal FTIR workstation with modular accessories for NIR, Raman, TGA-IR and GC-IR. Conduct fast bulk analyses with a built-in ATR in addition to the sampling compartment. One Touch ABX automatically exchanges optics to go from near-IR to mid-IR to far-IR on one spectrometer.
Learn more
How can you solve your toughest polymer challenges with only one click?
Monitor product consistency, troubleshoot, or deformulate complex materials and stay compliant. The Thermo Scientific Nicolet iS10 Spectrometer is a workhorse FTIR for regulated, fast-paced quality control, and analytical support laboratories.
Learn more
Need to expand your FTIR spectometer capabilities to include microsampling?
Rapidly identify materials with point-and-shoot simplicity using the Thermo Scientific Nicolet iN5 FTIR Microscope attached to your Nicolet iS10 or iS50 FTIR Spectrometer. Ensure product quality by visually screening for small contaminants and defects. Thermo Scientific OMNIC Spectral Analysis Software produces reliable answers that both a novice and expert can trust.
Learn more
You want to improve the speed and sensitivity of routine single point microspectroscopy measurements?
Rapidly acquire and analyze chemical images with the Thermo Scientific Nicolet iN10 MX Infrared Imaging Microscope to enhance your understanding of the chemical distribution of materials in heterogeneous samples. Incorporating a user-interface similar to an electron microscope, imaging software achieves consistent and reliable results and is designed for profession with any lever of experience in microscopy or spectroscopy.
Learn more
Need to visualize and compare chemical information in one sample?
Visually identify unknown particulates without being a spectroscopy expert using the intuitive Thermo Scientific DXR2xi Raman Imaging Microscope. Quickly analyze large areas with ultrafast chemical mapping. Real-time chemical imaging visually shows API distribution, polymorphs and crystalline structures. Visual software easily identifies multiple features to compare and contrast contaminants.
Learn more
Need to compound ISO/ASTM-compliant samples with only a small amount of material?
The Thermo Scientific HAAKE MiniJet Pro Sample Preparation System helps you to produce ISO / ASTM-compliant testing samples even from minimal amount of available materials. Work more efficiently and save money by reducing the time it takes to create samples for analyses and reduce expensive testing materials. The compact HAAKE MiniJet Pro sample preparation system is ideal for use with small-scale compounders like the Thermo Scientific HAAKE MiniLab and Thermo Scientific Process 11. Increase your testing and scale-up effectiveness and simplify your workflow.
  • Bring test specimen production to the bench in you lab. With optional features for temperatures up to 420 °C and cooling option 
  • Meet ISO standards for sample testing
  • Run material testing on small sample sizes (3–15 g) for quality control, tensile and stress tests, reliability and repeatability
  • Simplify cleaning for quick turnaround times between formulations
  • Benefit from an integrated solution for (micro)compounding and sample preparation for rheometers or use it as a standalone solution
Learn more
Do you want to measure structural changes and assess rheological properties of materials in R&D?
The Thermo Scientific HAAKE MiniLab III Micro-Compounder combines the advantages of the existing instrument with an intuitive user interface for standalone operation using a tablet computer. An app is available for Android devices. With the tablet app customers are enabled to program and run a defined measuring job routine and can monitor relevant process parameters such as torque, pressure, temperature and screw speed.
The HAAKE MiniLab III is used to measure structural changes during compounding and assess rheological properties of test materials ideal for R&D. The pneumatic force feeder guarantees easy sample loading. An integrated, bypass valve allows to recirculate the sample in a slit-capillary backflow channel for a defined time period or extrude it for further testing. The HAAKE MiniLab offers the additional advantages:
  • With its hinged barrel and fast cooling capacity, allows the freezing of the compound for further examination.
  • Measures relative viscosity during the compounding process 
  • Uses as little as 5 g or 7 ml of material 
  • Fits in a fume hood to limit exposure to hazardous materials 
  • Use as a standalone unit or as a computer-controlled system for rheological measurements
Learn more
You want to produce novel polymer compounds with a range of 20 grams per hour to 2.5 kg per hour?
The Thermo Scientific PROCESS 11 compact bench-top extruder helps you compounding in a range of 20 grams per hour to 2.5 kg per hour. Benefits that ease your daily work are for instance:
  • Easy operation by intuitive touch screen control 
  • Integrated feeder control 
  • Fast cleaning due to clamp shell barrel design with removable top half barrel
The process parameters obtained with PROCESS 11 are directly scalable with all our larger scale extruders such as Thermo Scientific EuroLab 16 and Thermo Scientific TSE MC 24.
A flexible sheet die and chilled sheet take off are available:
Sheet Die
  • 30 mm width with flexible lip height (max. 3mm) 
  • External, precise heating up to 350°C 
Chilled Sheet Take Off
  • 2- or 3-roll setup, easy to interchange 
  • Self-adjusting wind off for diameter compensation
Learn more
Understand melt behavior, temperature stability or influence of additives during product development
The Thermo Scientific HAAKE PolyLab OS Modular Torque Rheometer provides process-relevant material data like melt viscosity and behavior, temperature and shear stability. A variety of processing equipment including single screw and twin-screw extruders and measuring mixers are offered as accessories for the HAAKE PolyLab OS system. Furthermore, process modeling activities such as compounding polymers with additives, scale-up extrusion processes, using different L/D ratios to extrapolate to your manufacturing process line, simulation of production processes starting with 50 g of materials only and a wide range of post-extrusion accessories are available.
Consequently, you can optimize your process parameters, create test specimens or perform quality control on incoming and outgoing goods with regards to viscosity, dispersion or plasticizing behavior.
Learn more
Get the full picture and check out our comprehensive compounding solutions!
Check out our informative compounding web page featuring a number of valuable resources covering material development and analytics. Learn how our compounding solutions deliver efficient and cost-effective solutions to materials development and processing.
Learn more
Interested in rheological characterization of polymers or composites?
Do you need to test the rheological properties of your materials following the compounding process? Thermo Scientific HAAKE MARS rheometers measure the viscoelastic properties of polymer melts and solutions and solid samples. Test in shear, oscillation and elongation mode as a function of strain, frequency, time, or temperature.
The HAAKE MARS is a modular rheometer platform for individual demands and has much more to offer:
  • Technology advances such as patented technologies for air bearing and normal force sensor enabling accurate measurements
  • Well-conceived design for ease of use from quick connection of accessories up to measuring and evaluation software for beginners and advanced users 
  • A broad range of accessories enables users to customize solutions and tailor rheometer configurations
  • Combined methods for extended measuring capabilities including FTIR, microscopy, Raman, and DEA
Learn more
Would you like to understand rheological properties on a molecular level?
Discover the Rheonaut and learn more about the unique combination of the Thermo Scientific HAAKE MARS Rheometer with an FTIR Spectrometer!
Learn more
How can an individual, intuitive and intelligent rheometer simplify your quality control?
Discover the capabilities of the Thermo Scientific HAAKE Viscotester iQ rheometer. Established as the quality control rheometer in many industries. A rheometer with unique oscillation mode capabilities in its class. Convince yourself. Seeing is believing.
Learn more
Do you need accurate thickness and basis weight data for your extruded films, coatings and packaging?
Extend the shelf life of food products and protect medical instruments from contaminants! The Thermo Scientific 21Plus HD Measurement and Control System measures the basis weight or thickness of extruded films and coatings, ensuring uniform quality product across the width of the web, while reducing waste and precisely controlling raw material use! The 21Plus HD is the latest generation of this flagship quality control system and is the perfect solution for film extrusion, extrusion coating, flexible and aseptic packaging and technical films. Combined with the system's multilayer blown film application, it is also ideal for producing barrier layer films for the food, medical, and specialty film markets.
  • Comprehensive, customizable measurement and control platform
  • Supports multiple frames and sensors
  • Ensures improved product quality, process efficiency and raw material savings
  • Robust performance and easy to maintain 
  • Fast return on investment 
New generation system highlights:
  • New widescreen format incorporating full HD resolution
  • Improved navigation layout with minimal learning curve
  • Full range of functionality supporting all current applications
  • New 27” & 23” wide aspect touchscreen monitors
Learn more
Do you need superior measurement and control for your plastic films, coatings and packaging?
Manufacturing process control and repeatability is key to ensuring your products have superior quality, meet industry standards and are attractive to consumers. Producers of plastic films, extrusion coatings and blown films can get accurate thickness and moisture data, improve product quality, and precisely control raw material use with an easily-calibrated infrared thickness sensor. Whether you need a coating thickness gauge, paint thickness gauge, or film thickness gauge, the Thermo Scientific PROSIS Infrared Process Analysis Thickness Sensor uses the full spectral response of the near infrared range to analyze materials on the line, providing precise, multi-component thickness data that improves production quality and reduces waste. With a range of new standard calibrations, Prosis now measures an extensive range of materials out of the box with no further calibration effort. For all other materials EZCalPlus, an offline calibration tool, or SnapCal , a simple online calibration tool can be used.
  • Easy to use and calibrate
  • New standard calibrations for PP / PET / PE / PC / PA / PI
  • Simple online or offline calibration tools support all other materials
  • Exceptional performance for both single and multi-layer product structure
  • Completely safe non-nuclear solution - no license required
  • Low cost of ownership
Learn more
We hope to see you at the show!
Be inspired at Booth C106 in Hall 11 by how our innovative solutions can accelerate your everyday analytics!
Request information or schedule an appointment here