Webinar
May 03, 2017 - May 03, 2017
Polymer pellets
Combining rheology and Raman spectroscopy provides comprehensive insight material behavior. Rheology is the perfect technique to correlate absolute flow and deformation characteristics, but doesn’t provide insights into what is happening at the molecular level when rheological changes occur. That’s where Raman comes in. Raman is an effective and non-invasive method that is sensitive to structural composition of materials; providing the capability to differentiate between a material’s molten and crystalline forms. 
When these two techniques work together you can see a direct correlation of molecular composition and structure to the mechanical properties. Here are other benefits for multi-modal analysis with a rheometer and Raman spectrometer: 
  • Studying phase transition behavior
  • Insight for in-situ characterization and monitoring
  • Advantage of maximizing and synchronizing the information gathered from a single measurement
  • Observation of transient information during fast processing conditions
This webinar presents the results from the combination of a Thermo Scientific™ HAAKE™ MARS Rheometer with a Thermo Scientific™ iXR™ Raman Spectrometer. 
Register to watch this on demand.