Greater flexibility. Trusted results.
Greater flexibility. Trusted results.
Achieve true minimum inhibitory concentration results using manual susceptibility plates which offer modular, customizable testing options to accommodate laboratories of all sizes. Thermo Scientific™ Sensititre™ plates can be inoculated with our multichannel pipette or automated inoculation delivery system, then read manually with either a mirrored viewbox or using the Thermo Scientific™ Sensititre™ Vizion™ Digital MIC Viewing System.  
Looking for Sensititre plates for automated reading?
Featured products
Sensititre™ Gram Negative Urine Plate
Sensititre™ Gram Positive MIC Plate
Sensititre™ YeastOne™ Plate w/ Anidulafungin and Micafungin
Plate types
Thermo Scientific™ Sensititre™ 96-well microtitre plates are available in both standard and custom formats, with in-vitro diagnostic (IVD) or research use only (RUO) markings. With over 240 antimicrobials available, choose the plates to meet your unique needs.
Related products
Evaluate susceptibility MIC plates
Review this evaluation of Sensititre susceptibility MIC plates for the MIC determination of gram negative isolates for Amoxicillin, Amoxicillin/Clavulanic Acid, Cefotaxime, Meropenem and Piperacillin-Tazobactam.
 Download the poster
Featured videos
Join the fight against antibiotic resistance
Learn about the importance of antimicrobial stewardship policies and practices and effective implementation. 
 View slides and certificate ›
Professor Dilip Nathwani answers your questions ›
Sensititre introductory video
Sensititre system workflow video
Resources