Applied Biosystems® instruments have a proud history of leading the biotechnology industry. Inventors of fluorescence-based automated sequencing technology as well as real-time PCR science, we are now among the world’s leading companies pioneering next-generation sequencing as well as numerous other innovations. Unless otherwise noted, all instruments are for research use only, not for use in diagnostic procedures.
Sequencing instruments
  3500 Genetic Analyzer
3500 Series Genetic Analyzers
One of the most user-friendly genetic analyzers on the market.
  • Snap-in-and-run, preformulated consumables packaging
  • Auditable tracking of instrument performance and consumables usage
Personal Genome Machine™ (PGM™) Sequencer
Single day workflow from sample to annotated variants for gene panel sequencing featuring Ion AmpliSeq™ target technology, Ion PGM™ System, and the upcoming, automated Ion Chef™ System.**
  • Most accurate for multiple gene panels
  • Lowest cost to buy and run
Ion Proton™ System
The Ion Proton™ System removes the high cost and complexity of today's genome-scale sequencing, bringing it to your lab, on your budget, and on your schedule.
  • End-to-end workflows for rapid, high-quality sequencing results
  • Ideal for exome and transcriptome sequencing
Real-time PCR instruments
Instrument/ Capacity
96-well plates, 0.1 mL tubes
48-well plates, 0.1 mL tubes
7500 Fast
96-well optical Fast plates.
ViiA™ 7
TaqMan® Array Microfluidic Cards
QuantStudio™ 12K Flex Real-Time PCR System
(10–100 μL reactions),
Fast 96-well
(15–30 μL reactions)
(5–20 μL reactions)
TaqMan® Array Cards
(~1 μL reactions)
OpenArray® Plates
(33 nL reactions)
Temperature Uniformity± 0.5°C within 30 seconds of clock start± 0.5°C within 30 seconds of clock start±0.5°C, within 30 sec of reaching temperature set point±0.25°C (±0.75°C for OpenArray® block)
Fast blockYes
7500 & 7500 for HID
(96-well and 384-well)
Performance Specifications
Demonstrated Sensitivity10 copies10 copiesDown to 1 copyDown to 1 copy
Dynamic Range9 logs of linear dynamic range5 logs guaranteed, 9 logs demonstratedUp to 9 logs of linear dynamic rangeUp to 9 logs of linear dynamic range (up to 7 logs for OpenArray® block)
Instrument Specifications
(W x D x H)
W 23.6 cm
D 51.2 cm
H 42.7 cm
W 34 cm
D 45 cm
H 49 cm
W 53.3 cm
D 63.5 cm
H 64.5 cm
W 50.5 cm
D 67.2 cm
H 73.8 cm
Multiplex QuantitationYesYesYesYes (on all non-OpenArray® blocks, duplex on OpenArray®)