Accurate, reproducible results in immunoassay, sample prep and R&D testing
Accurate, reproducible results in immunoassay, sample prep and R&D testing
Achieve precise performance and accurate results in specialized research and industrial applications with Thermo Scientific™ beads for optimizing assays, for analyzing particulate materials of various sizes and for testing smoke detectors and filtration media.
View all products
Key Applications
  • Opti-Link: Covalent couping of proteins
  • Opti-Bind: Turbidimetric assays, slide agglutination tests
  • Power-Bind: Clinical diagnostic kit development, nucleic acid research
  • 5000 and 7000 Series: Filter evaluation, fluid mechanics research, dispersion studies, light scattering and other R&D projects
  • Smoke-Check: Monitor transport time of air sampling smoke detectors
Featured products
5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions
7000 Series Copolymer Microsphere Suspensions
Opti-Bind™ Polystyrene Sulfate Particles
Resources