molecular-medicine.par.30588.image.0.0.1
The final part of any genetic analysis assay is the analysis and reporting.We offer a suite of software tools to assist with your laboratory’s goals. Unless otherwise noted, all software tools are for research use only, not for use in diagnostic procedures.
EDCNet
EDCNet (Electronic Data Collection—Internet-based reporting) provides subscribers with the ability to directly enter and then graph data generated from quality control (QC) results used to monitor serological or nucleic acid tests.
  • Customizable for your laboratory’s testing protocol
  • Includes scatter graphs and bar graphs for assay performance valuation and result comparison between labs
  • Includes Shewart (Levy-Jennings) charts that facilitate the early detection of QC result trends
Learn more
TaqMan® Genotyper™ Software
A free SNP genotyping data analysis tool for use with TaqMan® SNP Genotyping Assays.
  • State-of-the-art genotype-calling algorithm with an intuitive user interface
  • Enhanced multiplate analysis features to meet the requirements of emerging markets and future research
Learn more
CopyCaller® Software
Free software developed specifically for TaqMan® Copy Number Assay data analysis.
  • Easy-to-use software with a graphical interface enables quick copy-number calculations
  • Assigns confidence values for each copy-number call and includes outlier removal function
Learn more
SeqScape® Software
Reference-based sequence analysis for mutation detection, SNP discovery, allele identification, sequence confirmation,
and more.
  • Easy-to-use tool for large-volume data analysis
  • Offers basecalling quality values for all bases, consensus, and mutations
Learn more
Variant Reporter® Software
Designed for reference-based and nonreference-based analysis such as mutation detection and analysis, SNP discovery, and sequence confirmation.
  • Designed for basic and clinical researchers, for genotyping projects and secondary analysis
  • Automates variant detection, saving time and money
30 Day Demo Initial License
GeneMapper® Software
GeneMapper is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Applied Biosystems® electrophoresis-based genotyping systems.
Learn more
LifeScope™ Software
SOLiD® LifeScope™ Software uses a flexible pipeline architecture to provide maximum flexibility and easy of use for performing high-throughput data analysis for SOLiD® instrument data.
  • Streamlined interface—command line and simple web-interface options provide flexibility for user preference
  • Efficient infrastructure—designed towork on offline clusters and cloud computing configurations to relieve instrument bottlenecks
  • Consolidated analysis—standard base sequence file exports enable the combination of data from multiple sequencing platforms for single project-based analysis
  • Versatile application—includes mapping, pairing, SNP finding, structural variations, and whole transcriptome analyses
Learn more
DataAssist™ Software
DataAssist™ Software is a free, easy-to-use data analysis tool that utilizes the comparative Ct (ΔΔCt) method to rapidly and accurately quantitate relative gene expression across a large number of genes and samples. You can import raw data from hundreds of plates or TaqMan® arrays, and change analysis settings including selection of normalization controls and method (single or multiple control genes). It provides instant responses and fast calculations, allows normalization using multiple reference genes, and provides analysis results in content-rich tables and scalable graphic charts that are easily exported.
Learn more
RealTime StatMiner® Software
Advanced data mining software for real-time PCR data analysis. RealTime StatMiner® Software provides a high-powered, straightforward solution to data analysis for qPCR.
Learn more