probe-primer-analyte-specific-reagents-showcase-image-230x195
Confidence for your test development
Applied Biosystems™ MGB probe and unlabeled primer analyte specific reagents (ASRs) are manufactured to help meet requirements for test development. Applied Biosystems Probe and Primer ASRs can be used across a wide variety of applications, including gene expression, genotyping, and copy number analysis.
Applied Biosystems Probe and Primer ASRs are manufactured in a registered facility in accordance with current Good Manufacturing Practices (cGMP) with a quality management system certified to ISO 13485:2003.
Download flyer ›
Specifications
MGB Probe ASR
Reporter dyes
FAM, VIC, NED
Normalized final yield
50 or 100 nmol
Format
In solution
Length
11–35 bases
Purity
≥85%
Unlabeled Primer ASR
Normalized final yield
100 nmol
Format
Dried down or in solution
Length
11–35 bases
Purity
≥85%
Applied Biosystems Probe and Primer ASRs are all HPLC-purified.
Analyte Specific Reagent. Analytical and performance characteristics are not established.
How to order
To order Applied Biosystems Probe and Primer ASRs, send an email to GMP-Oligos@thermofisher.com to initiate the process.
Once the order is placed, we typically ship in 3–4 weeks, but turnaround time can vary depending on order size. A printed Certificate of Analysis will ship with your order; additional copies can be provided upon request. 
Applied Biosystems Probe and Primer Analyte Specific Reagents
Cat. No.
Name
Reporter dye
Final yield
Format
Length
A26930
MGB Probe
FAM
50 nmol
In solution
11-35 bases
A26924
100 nmol
A26929
MGB Probe
VIC
50 nmol
In solution
11-35 bases
A26923
100 nmol
A26926
MGB Probe
NED
50 nmol
In solution
11-35 bases
A26922
100 nmol
A26935
Primer (Dry)
-
100 nmol
Dried down
11-35 bases
A26934
Primer (TE)
-
100 nmol
In solution
11-35 bases
* These products are currently available for sale in the US only. 
Analyte Specific Reagent.  Analytical and performance characteristics are not established.
Visit the RUO probes and primers page ›