Convenient, accurate nucleic acid separation
  • Great time saver, no hassle system
  • High QC standards and reproducibility
  • Reduced staff training, with no buffer preparation, pouring, and disposal issues
  • No Ethidium bromide exposure
  • E-Editor™ 2.0 software provides results in an easy format to interpret and document
E-Gel®  are bufferless, pre-cast agarose gels designed for fast,
high-throughput DNA electrophoresis.
The E-Editor 2.0 software effectively reconfigures you E-Gel images.
E-Gel® frees up space, routine work time, disposal concerns, and increases safety.
Ordering Information
Sku Product Catalog Size Price
Quantity
A10570 E-Gel® 96 (2% agarose Ethidium Bromide)
8 pack
A10571 E-Gel® 48 (2% agarose Ethidium Bromide)
8 pack
A10572 Mother Base 1    
A10573 Daughter Base 1    
A10572 UK Mother Base UK plugs 1    
A10572 EU Mother Base EU plugs 1