showcase.par.43404.image.0.0.1
The uTYPE® Dx Sequencing Analysis Software combines rapid data analysis with effective results management. Interpret data from a single user interface and navigate easily through key positions. Utilize single-click editing without compromising your original data file. A unique filter function eliminates mismatched allele combinations from consideration, while customizable reports containing interpreted results are available for convenient review.
Updates to data libraries are available quarterly.
HiRes Me Wizard
The uTYPE® Dx Sequencing Analysis Software HiRes Me Wizard leads you through sample analysis to the highest resolution possible, including systematic elimination of:
  • single mismatch combinations
  • null alleles
  • intronic polymorphism