PG1456-PJ6916-WE219192-Update-hero-banner-on-instruments-page-to-align-with-clinical-creative-Americas-F
Supporting your lab’s assay development needs along with a workflow to match your diagnostic requirements
Ion PGM Dx
The Ion PGM™ Dx System delivers the speed and simplicity of the Ion Torrent™ platform, combined with key workflow advantages designed specifically for regulated laboratory environments and in vitro diagnostic applications. Now with combined functions software* to enable maximum utility for clinical research.
Veriti Dx Thermal Cycler
The QuantStudio™ Dx Real-Time PCR Instrument offers your clinical laboratory performance you can rely on in a Fast 96-well diagnostic platform, as well as provides the flexibility your lab needs to grow with additional formats. The QuantStudio Dx instrument can be operated in either IVD mode (using the Quidel™ Molecular Direct C. difficile Assay) or Test Development mode (for assay development).
500 Dx Genetic Analyzers CS2 Instruments
3500 Dx Genetic Analyzers CS2 provide laboratories with reliable, versatile, and innovative tools for molecular diagnostic testing. Based on Sanger sequencing, one of the most widely used, widely trusted sequencing methodologies available, the 3500 Dx Genetic Analyzer CS2 can be operated in either IVD mode (for diagnostic HLA typing using SeCore® HLA typing kits) or RUO mode (to develop other applications).
The Applied Biosystems® 7500 Fast Dx Real-Time PCR
The Applied Biosystems 7500 Fast Dx Real-Time PCR Instrument is designed to deliver the performance required for high-quality results. Accommodating both 96-well plates for easier setup and tube strips for lower-throughput runs, the instrument enables assay development and validation projects on the same in vitro diagnostic instrument.
Veriti Dx Thermal Cycler
The Veriti™ Dx PCR Thermal Cycler provides the reliability of Applied Biosystems™ 9600 and GeneAmp™ systems in an IVD instrument. It features a powerful color touch screen to simplify instrument setup and provides precise control for PCR optimization, the ability to run six different thermal cycling profiles at once, and both Fast and standard PCR modes.
WE212319---Reagent-POD
WE212321---Trade-in-program-pod