E-Editor™ 2.0 software takes the digital image of your E-Gel® 48 or 96gel and reconfigures the lanes into one aligned image for fast and easy positive lane identification and documentation (Figure 2).
The E-Editor™ 2.0 software is available free of charge with the purchase of E-Gel® 48 and 96 gels and related equipment:
  • Works on all Windows® platforms
  • Recreates ‘virtual gel’ from the 96 format
  • Allows tracing back from well coordinates to lanes
  • Able to transpose vertical loading to horizontal interpretation
Raw data from an SSP typing experiment run on an E-Gel®
96 precast gel (A). The same gel after processing with the E-Editor™ 2.0 software (B).