For accurate, robust, reproducible SSP typing kits, we offer SSP UniTray® worldwide. We offer an extensive product line that encompasses over 50 kit formats for low and high resolution of HLA Class I and Class II.
  • High degrees of resolution
  • Simple post-amplification analysis
  • Cost-effective alternative to other molecular methods
  • Low and high resolution typing with one method
  • Frequently updated for new alleles
  • Superior technical and customer service
  • One protocol for all kits
  • Ready to use, pre-aliquoted PCR Buffer
  • Worksheet for manual analysis
  • UniMatch® PLUS software for easy interpretation
  • AmbiSolv® primer mixes for resolving ambiguities
  • Allele updating available on the website