The Prodigy™ Instrument will free up valuable technologists time and improve laboratory efficiency by automating the entire SSO assay. Run time is only 1.5 to 3 hours, including data collection. Its complete automation will give you superior reproducibility. The Prodigy Instrument’s Touchscreen will walk you through the easy assay set-up by providing step by step instructions along with your run’s specific reagent needs. 
  • Optimized Small Footprint: 27”W x 23”L x 23”H
  • User Friendly TouchScreen for machine set-up
  • Integrated camera takes a snapshot of the chip and transfers to software analysis for immediate interpretation
  • Microarray has 5x the capacity of today’s bead
  • Barcode reader to identify strip lots for traceability
  • Single and Dual hit Probes are designed for all known Exon 2 & 3 polymorphisms
  • Ability to run 1-12 strips (8-96) samples of any loci at a time
  • Identical 1.5 hour amplification strategy for all loci
*This product may be covered by one or more Limited Use Label Licenses (see Invitrogen catalog or www.invitrogen.com). By use of this product you accept the terms and conditions of all Limited Use Label Licenses. For Research Use Only. Not intended for any animal or human therapeutic or diagnostic, unless otherwise stated.