Get started using these apps by first accessing Thermo Fisher Cloud. Don't have an account? Start here ›
Jump to:     Real-time qPCR    CE Sanger sequencing    CE fragment analysis    Next-generation sequencing    Endpoint PCR    Synthetic biology    Mass spectrometry    Pipetting     Virology    Microbiology    Molecular spectroscopy    Protein biology    Digital PCR    Utilities    Oncology
apps-da
Design and Analysis
The Design and Analysis app offers the ability to create, edit, and analyze qPCR instrument files.
GT1
Genotyping
The Genotyping app includes improved visuals and integrated traces of allelic discrimination plots to allow thorough QC of SNP assays to accurately reflect the true signals versus background noise. 
HRM icon
High Resolution Melt (HRM) Analysis
This High Resolution Melt Analysis (HRM) app is designed for post-PCR analysis to identify variation in nucleic acid sequences. The method is based on detecting small differences in PCR melting (dissociation) curves. It is enabled by high-brightness, dsDNA-binding dyes used in conjunction with real-time PCR instrumentation that has precise temperature ramp control, advanced data capture capabilities, and access to software designed specifically for HRM analysis.
PA icon
hPSC Scorecard Analysis
The hPSC Scorecard Analysis app scores gene expression profiles generated using the TaqMan hPSC Scorecard Panel compared to a reference set of well-characterized pluripotent stem cell lines. It provides raw data as well as box plots, heat maps, and correlation plots for export.
PA icon
Presence/Absence Analysis
The Presence Absence Analysis app analyzes and interprets real-time PCR data or post-read data to determine if a specific target sequence is present (presence) or not present (absence) in a sample. It provides an easy-to-view presence/absence result in a plate grid view.
RQ1
Relative Quantification
The Relative Quantification app allows fast and powerful gene expression analysis with enhanced visual capabilities for relative quantification including integrated correlation and volcano and cluster analysis with the ability to drill down to amplification plots.
apps-sc
Standard Curve
The Standard Curve app offers reliable quantification of unknown quantities of genes and enables importing of standard curves from other experiments, providing analysis flexibility.
Back to top
QC
Quality Check
The Quality Check app automatically checks CE sequence trace quality. It provides a results summary based on quality parameter settings and auto-flags lower quality traces for further inspection. Users can quickly and easily navigate to questionable or borderline data, and analyze, make adjustments, or exclude traces from the study.
Variant Analysis
The Variant Analysis app finds variants in samples sequenced on Applied Biosystems genetic analyzers. It reports variants at genomic coordinates and allows users to export variant calls in standard vcf format. It also reports genomic annotations for SNPs and provides links to external databases.
Learn more ›
Next-Generation Confirmation
The Next-Generation Confirmation app confirms NGS variants using CE technology. It allows users to easily visualize the variants detected by both NGS and CE platforms and to export confirmed variants in standard vcf format.
Learn more ›
SeqStudio Plate Manager 
The SeqStudio Plate Manager app enables the user to set up plates and configure different application-type runs for the Applied Biosystems SeqStudio Genetic Analyzer. It also supports remote monitoring and remote control of runs.
Back to top
CE fragment analysis
Peak Scanner
Peak Scanner
The Peak Scanner app offers peak identification and fragment sizing for application-specific capillary electrophoresis assays.
MSA
New Microsatellite Analysis
The Microsatellite Analysis app is microsatellite genotyping software that allows you to analyze a mixture of DNA fragments, separated by size. This analysis provides a profile of the separation, precisely calculates the sizes of the fragments, and determines the microsatellite alleles present in the sample. Microsatellite analysis is commonly used to study Microsatellite Instability in cancer and Triplet Repeat Expansion in neurodegenerative diseases, and for Species Identification & Characterization and Human Sample Authentication.
Back to top
Next-generation sequencing
Ion Reporter
Ion Reporter Software provides a suite of data analysis tools for variant detection, annotation and reporting for the Ion PGM, Ion Proton, and Ion S5 and Ion S5 XL Systems. The software identifies variants such as SNPs, indels, CNVs, and gene fusions, using pre-configured and customizable workflows optimized for Ion AmpliSeq panels. Using annotations from >20 public databases, including COSMIC, dbSNP, and OMIM, Ion Reporter Software has advanced filtering capabilities to focus on relevant variants. Visualization and reporting capabilities allow for sample comparison and creation of interpretive research reports for selected variants. Also, identify complementary CE and TaqMan Assays for variants of interest.
Learn more ›
Endpoint PCR
PD
PCR Designer
The PCR Designer module offers the ability to create and edit thermal protocols for SimpliAmp & ProFlex endpoint PCR instruments.
Back to top
Synthetic biology
apps-CR
CRISPR Search and Design Tool
Use the CRISPR app to search our database of >600,000 predesigned CRISPR gRNAs or to analyze your sequence of interest for de novo gRNA designs for use in genome editing with CRISPR-Cas9 technology. The CRISPR module enables easy purchase of CRISPR-Cas9 and associated Thermo Fisher Scientific products.
GQ
GeneArt QA
Use this module to browse, download and share Quality Assurance Documentation (QAD) for all your orders from GeneArt. Note that only QAD for orders shipped after you confirmed the Terms Of Use for the service will be uploaded and made available for review.
GSA
GeneArt™ Strings™ Assistant
The GeneArt™ Strings™ Assistant makes it easy to order your DNA sequences as GeneArt Strings DNA fragments.
ICD
Invitrogen™ CRISPR Array Designer 
Use The Invitrogen™ CRISPR Array Designer Tool to design large number of CRISPRs for cell engineering experiment(s). The software will also assist customers in arranging oligonucleotides (containing CRISPR guide-RNAs) on 96-well plates. This is not associated with any instrument but has the underlying algorithm used by the GeneArt CRISPR Search and Design Tool. All of these products can be ordered by customers from Thermo Fisher Scientific.
OP
Oligo Perfect 
Use the OligoPerfect™ designer to design oligonucleotide primers for your PCR-dependent experiments. In addition to designing primers, the OligoPerfect™ designer enables easy purchase of oligonucleotides by Thermo Fisher Scientific.
SV
Sequence Viewer 
The Sequence Viewer module will allow users to load DNA sequence files and view relevant molecule information visually. Molecule features can be viewed as annotated sequences in intuitive graphical and sequence maps.
TAL
TAL
Use the TAL module to search our database of >600,000 predesigned TALs or to analyze your sequence of interest for de novo TAL designs for use in genome editing with TAL technology. The TAL module enables easy purchase of TALs and associated products by Thermo Fisher Scientific.
Back to top
Mass spectrometry
PO Icon
Pathway Over-Representation Analysis
The Pathway Over-Representation Analysis app uses statistical analysis to detect biological pathways that are either over- or under-represented in a list of input proteins. The software accepts a list of protein identifiers as input and reports significant WikiPathways and optionally KEGG (Kyoto Encyclopedia of Genes and Genomes) pathways associated with the list.
PO Icon
Sample Profiler
Sample Profiler identifies unknown compounds in high resolution accurate mass spectrometry sample data derived from defined populations. Once identified, these unknown compounds are compared against a database of representative compound profiles, complete with their chromatographic peak ranges. Data differences are analyzed via integrated hypothesis testing tools. Using this process, Sample Profiler elucidates how a new “challenge” sample differs from an existing representative profile; the app also measures how a sample population profile changes with time, location, production method, and more.
PO Icon
MS Instrument Connect
MS Instrument Connect provides the ability to monitor your LC-MS and GC-MS analyses remotely from any web-enabled device. Use this application to monitor real-time mass spectrometer status, track system utilization and historical events, and create email notifications to keep you informed when analytical runs are complete or when system errors occur. MS Instrument Connect is integrated with the Instrument Connect App, allowing you to track your systems from any iOS or Android device.
PO Icon
Omics Comparator
The Omics Comparator analysis software generates Venn diagrams to compare up to 5 lists of input proteins. The analysis can also work on lists of genes. The software displays set membership together with molecular annotations including color coded Gene Ontology terms.
Pipetting
Back to top
PO Icon
My Pipette™ Connect Utility
The My Pipette Connect Utility for Windows enables E1-ClipTip electronic pipettes to link with the My Pipette Creator app. Please download and install to get started. The My Pipette Creator app helps to increase productivity by leveraging the power of the Thermo Fisher Cloud and the connectivity feature of the E1-ClipTip electronic pipette. This app allows users to efficiently create pipetting programs, collaborate by sharing programs with colleagues, and update pipette firmware. Programs can be easily transferred to and from the E1-ClipTip wirelessly or via a USB connection. Additionally, pre-written and validated protocols designed specifically for Thermo Fisher assays and kits are available which can dramatically reduce experiment setup time and minimize potential errors. For more information, please visit thermofisher.com/mypipette
PO Icon
My Pipette™ Creator 
The My Pipette Creator app helps to increase productivity by leveraging the power of the Thermo Fisher Cloud and the connectivity feature of the E1-ClipTip electronic pipette. This app allows users to efficiently create pipetting programs, collaborate by sharing programs with colleagues, and update pipette firmware. Programs can be easily transferred to and from the E1-ClipTip wirelessly or via a USB connection. Additionally, pre-written and validated protocols designed specifically for Thermo Fisher assays and kits are available which can dramatically reduce experiment setup time and minimize potential errors. For more information, please visit thermofisher.com/mypipette
Back to top
Virology
PO Icon
ViroScore®-HIV
The HIV Sanger software & database solution to perform genotyping and subtyping analyses, and measure potential impact on disease probability through more than 7 regularly updated algorithms, organized into easy-to-use reports.
Microbiology
Back to top
PO Icon
CosmosID
CosmosID's platform identifies bacteria and viruses in a whole genome shotgun metagenomic sample and is able to discriminate pathogens from their ‘near neighbors’ using statistical and computational methods, with no prior assumptions as to what is present in the sample.
Molecular spectroscopy
ND
NanoDrop 
The NanoDrop app gives you the flexibility to view and work with dsDNA, ssDNA and RNA data acquired with the NanoDrop One and the NanoDrop 2000 instruments on any device. You can access all of the Acclaro Sample Intelligence Technology information associated with your NanoDrop One data. Contaminant information can be displayed and reviewed with an interactive spectral display.
OA
OMNIC™ Anywhere
The OMNIC™ Anywhere app allows you to extract the content of an SPA file and visualize its data within an interactive spectral display. You can perform basic operations on it, such as find the peaks of the spectrum trace and download its graph view as an image ready to be included in your reports.
Back to top
Protein biology
IBA
iBright Image Analysis Software
iBright Image Analysis (iBA) software is an easy to use application for automated data analysis and management of digital images depicting electrophoresis gels and blots. The software can be used for editing images (e.g., to adjust brightness/contrast, rotation, crop, false coloring, etc) as well as identification and quantitation of bands, background subtraction, relative and absolute quantitation, molecular weight analysis, and report generation. This application supports the iBright CL 1000 and iBright FL 1000 Imaging systems.
PQ
ProQuantum
The ProQuantum™ software enables you to analyze ProQuantum™ protein immunoassay results acquired from any qPCR instrument. The analysis features include standard curve qualification with built-in outlier detection, interpolation of unknown samples for quantitation, and group-wise statistical analysis. Step-by-step guidance is provided for preparation of an assay run, including setting up the standard curve, plate layout design, and customized bench instructions.
Back to top
Digital PCR
AS
AnalysisSuite 
This application is for the analysis of dPCR data from the QuantStudio 3D Instrument platform. The software yields absolute Copies/uL and Target/Total results.
Utilities
Cloud Connect Utility
Cloud Connect Utility is a desktop client for the Thermo Fisher Cloud (TFC) user to continuously synchronize/upload data to their TFC account and view realtime upload status. CCU lets you securely upload your data files generated from instruments that aren’t cloud-enabled, such as the Applied Biosystems™ StepOne™, StepOnePlus™, 3500XL, QuantStudio6Flex, QuantStudio7Flex, QuantStudio12K Real time PCR, 7900 & 7500 real-time PCR systems, among others. You can also store and backup files, analyze your saved data using TFC’s web-based apps, or securely share your datasets with your colleagues. Windows and Mac versions available.
Download ›
More about the utility ›
 Instructions for installation and use ›
Education Connect
Education Connect is an online classroom companion supporting LabCoat Live™— the first of its kind, virtual hands-on learning program. The capability provides all of the resources needed for your individualized scientific learning plan. Features include live lecture space for theoretical and data analysis overviews, resource libraries for course lecture notes, protocols, instructional videos, and a discussion board enabling peer-to-peer and participant-to-instructor communication for up to two months after course completion.
Back to top
Oncology
Oncomine Knowledgebase Reporter 
Oncomine Knowledgebase Reporter helps you prioritize variants, find the relevant cancer drivers, and create a report that enables contextual investigation of sample-specific variants to understand their use with respect to current labels, guidelines, and global clinical trials. It is a genomic analysis tool developed specifically for further examination of NGS data, enabling streamlined access to a final report in just a few easy steps.