USDA-licensed, real-time PCR screening and subtyping kits for the rapid detection of swine influenza virus (SIV)
Thermo Fisher Scientific animal health products have been at the forefront of recent advances in production animal diagnostics, and supply some of the leading modern diagnostic tools to veterinary laboratories around the world.
In keeping with our commitment to deliver the highest quality next-generation molecular tools, we provide several diagnostic solutions licensed by the USDA. This includes our USDA-licensed VetMAX™-Gold SIV solutions: rapid, easy-to-use real-time PCR screening and subtyping kits that detect influenza virus in pigs with superior sensitivity, specificity, repeatability and reproducibility.
About SIV
SIV is a highly contagious viral infection of pigs caused by a variety of closely related influenza A viruses. These viruses are noted for their ability to create new strains such as H1 N1, H1 N2, and H3 N2. An outbreak is typically preceded by 1–3 individual cases and then spreads rapidly within a herd, mainly by aerosolization (coughing, nasal discharge) and pig-to-pig contact.
SIV causes a respiratory disease characterized by coughing, sneezing, nasal discharge, fever, lethargy, difficulty breathing, loss of appetite, and in some cases may contribute to reproductive problems and abortion. Although mortality tends to be low, morbidity can reach 100%, which can place an economic burden on producers due to treatment expenses and animals arriving late to market as a result of delayed weight gain.
Highly accurate diagnostic solutions for the rapid detection of SIV
The VetMAX™-Gold SIV Solutions are USDA-licensed diagnostic tests that have successfully passed the USDA’s stringent review process. This process requires demonstration of the effectiveness of the real-time PCR test and manufacturing of the product in a USDA-licensed and -inspected biologics facility.
Veterinary diagnosticians rely on our molecular reagents and instrumentation to detect and control disease. With our VetMAX™-Gold SIV product they have a USDA-licensed diagnostic test for SIV containing all the necessary reagents and controls in a single detection kit.
Confidence in results
Our USDA-licensed kits have demonstrated superior repeatability, reproducibility, and sensitivity. They target multiple genomic regions to help reduce the risk of false negatives that can be caused by genomic mutations.
Fast and simple to use
The simple, validated, and easily integrated workflow helps minimize sample handling and cross-contamination. It delivers a fast and cost-effective SIV detection method that yields results in as little as 1.5 hours.
Enables cost savings
Our USDA-licensed, real-time PCR kits help reduce the assay validation burden for laboratories and also helps decrease variability of results, allowing for reproducibility through assay standardization.
Confidence in results backed by data
VetMAX™-Gold SIV Detection Kit - A study was conducted of 226 nasal swab field samples (121 positive and 105 negative) of various SIV subtypes. The complete workflow was carried out by external collaborators as part of the USDA-licensing effort. The SIV status of each sample was determined prior to testing with virus isolation and/or sequencing. The assay produced 98.4% sensitivity and 99.1% specificity. The only discrepant positive samples were from very low-titer samples amplified near the assay’s limit of detection, which resulted in inconsistent detection upon retesting. The single discrepant negative sample was confirmed to be negative upon reisolation and repeated analysis by quantitative reverse-transcription PCR (qRTPCR).The original positive test was likely due to crosscontamination during isolation or qRT-PCR setup.
VetMAX™-Gold SIV Subtyping Kit - The kit produced 98.2% sensitivity and 100% specificity for identifying the SIV subtype from porcine nasal swab samples. This study indicates that RNA isolated from diagnostic porcine nasal swab samples, tested with the VetMAX™-Gold SIV Subtyping Kit in conjunction with the VetMAX™-Gold SIV Detection Kit, can provide an economical and rapid solution for SIV subtype identification.
Table 1. Results of SIV-positive sample sensitivity testing.
Call No. of samples
Final call: true positive 166
Final call: false negative 3
Diagnostic sensitivity: 98.2%
Table 2. Results of SIV-negative sample specificity testing.
Call No. of samples
Final call: true negative 150
Final call: false positive 0
Diagnostic specificity: 100%
VetMAX™-Gold SIV Solutions product flyer
Our vision is to provide comprehensive solutions to help diagnose farm animal health issues more quickly and accurately than ever before—and thereby enhance animal health and productivity. To learn more about our VetMAX™-Gold SIV solutions, please download our product flyer.
  See product flyer ›