Thermo Scientific moisture and elemental analyzers ensure consistent bulk raw material quality
Increase productivity, eliminate errors
Elemental analyzers provide coal producers with with real-time coal sampling and analysis to help them proactively address process variations to ensure more consistent coal blends and improved fuel quality. Cement producers strive for consistent raw material quality with minimal chemistry deviation to improve product quality, boost efficiency, and lower production costs.
Thermo Scientific™ online sampling and analysis systems integrate directly into a new or existing conveyor application and determine the elemental composition of all the raw materials being transported by conveyor, eliminating many of the errors and costs associated with material sampling. 
Contact a sales representative
Quick links
Featured sampling and analysis systems
Ramsey™ Model SWAX-7000 Primary Sweep Type Sampler
CQM Flex Coal Analyzer
CB Omni Fusion Online Elemental Analyzer
Sample directly from material on a conveyor belt with the Thermo Scientific Ramsey Model SWAX-7000 primary sweep type sampler
Procure a representative sample directly from material on a conveyor belt simply and economically with the Thermo Scientific Ramsey Model SWAX-7000 Primary Sweep Type Sampler. This sampler can be used as a stand-alone sampling device or as a primary in a multi-stage mechanical sampling system.
Obtain important data for blending and sorting of coal. The Thermo Scientific ECA Elemental Crossbelt Analyzer uses cutting-edge pulse processing technology to measure parameters such as ash, moisture, and sulfur.
Measure the major coal quality parameters in real time with the Thermo Scientific CQM FLEX Analyzer. The CQM FLEX is a full-featured analyzer that can incorporate either Prompt Gamma Neutron Activation (PGNAA) technology or Pulsed Fast Thermal Neutron Activation (PFTNA).
Reduce process upsets and increase mill throughput with the Thermo Scientific CB Omni Fusion Online Elemental Analyzer. The system provides minute-by-minute elemental analysis of the entire material volume using either PGNAA or PFTNA.
Get accurate, real-time, cost-effective measurements with the Thermo Scientific Low Frequency Microwave (LFM3) Moisture Analyzer. It can be easily integrated and installed in a variety of applications.
Featured sampling and analysis videos
Resources
Support
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.