hdi-9mm-screw-thread-vial-closures-230x195
Innovative closure technology. Unmatched performance.
The Thermo Scientific™ National Target™ DP™ 9mm C5000 series and the Thermo Scientific™ Chromacol™ K-Series vial closure systems are innovative, flexible and performance driven vial closure systems built on the Advance Vial Closure System (AVCS) technology platform.
Based on Advance Vial Closure System (AVCS) technology
Chromacol and National Target DP 9mm AVCS color closures help chromatographers achieve more reliable and efficient sample analysis. Designed to accommodate nearly all chromatography autosamplers fitted for 12 x 32cm vial trays, the Chromacol K-Series and National C5000 series closures effectively eliminate septa push-through and significantly enhance productivity and flexibility in the laboratory.
9mm Screw thread vial closure types
AVCS closures are offered in a larger assortment of cap colors and septum formulations than any other supplier providing unmatched flexibility in meeting laboratory needs. For easy visual identification of samples around the laboratory, select from a range of colors for use with clear, amber or plastic vials.
Features of 9mm Wide Opening, Integral Membrane and Solid Closures
  • Nearly eliminates septa push through due to AVCS technology
  • Increased sealing capability
  • Unlimited options in selecting the best septum for your instrument and applications
  • Cost efficient closure as current choices for resolving push through (bonded septa) become obsolete
  • Optimized ergonomics, fine texturing and evenly space ribbing for handling even with poorly fitting gloves
A wide opening vial deserves a wide opening closure, utilizing AVCS, providing greater area for injection needle penetration.
In addition to AVCS technology, the integral membrane provides a high value option for applications where the environment is more conducive to a polypropylene membrane.
AVCS technology provides maximum protection of samples during down to -80 °F.
National Target DP 9mm C5000 Series Vial Closure
Description Quantity Part Number
National Target DP Blue 9mm Screw thread Cap with Natural PTFE/Blue Soft Silicone Septum2 100/PK C5000-44B
National Target DP Blue 9mm Screw thread Cap with Natural PTFE/Blue Soft Silicone Septum, Pre-slit2 100/PK C5000-45B
National Target DP Blue 9mm Screw Thread Cap with Ivory PTFE/Red Rubber Septum 100/PK C5000-51B
National Target DP Blue 9mm Screw Thread Cap with Red PTFE/White Silicone/Red PTFE Septum 100/PK C5000-53B
National Target DP Blue 9mm Screw Thread Cap with Red PTFE/White Silicone Septum 100/PK C5000-54B
National Target DP Blue 9mm Screw Thread Cap with Blue PTFE/White Silicone Septum, Preslit 100/PK C5000-55B
National Target DP Clear 9mm Screw Thread Cap with Integral Polypropylene Membrane 100/PK C5000-50
National Target DP Blue 9mm Screw Thread Cap with Integral Polypropylene Membrane 100/PK C5000-50B
National Target DP Blue 9mm Screw Thread Closed Top Cap with Ivory PTFE/Red Rubber Septum 100/PK C5000-99B
National Convenience Kit, Clear Glass, Blue DP Cap w/ Ivory PTFE/Red Rubber Septum 100/PK C5000-80
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Ivory PTFE/Red Rubber Septum 100/PK C5000-80W
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Red PTFE/White Silicone/Red PTFE Septum 100/PK C5000-86W
National Convenience Kit, Clear Glass, Blue DP Cap w/ Red PTFE/White Silicone Septum 100/PK C5000-92
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Red PTFE/White Silicone Septum 100/PK C5000-92W
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Blue PTFE/White Silicone pre-slit Septum 100/PK C5000-95W
National Assembled Vial Kit, Clear Glass, Ivory PTFE/Red Rubber Septum, Blue DP Cap 100/PK C5000-180
National Assembled Vial Kit, Clear Glass w/I-D Patch, Ivory PTFE/Red Rubber Septum, Blue DP Cap 100/PK C5000-180W
National Assembled Vial Kit, Clear Glass w/I-D Patch, Red PTFE/White Silicone/Red PTFE Septum, Blue DP Cap 100/PK C5000-186W
National Assembled Vial Kit, Clear Glass w/I-D Patch, Red PTFE/White Silicone Septum, Blue DP Cap 100/PK C5000-192W
2Soft durometer septa are a effective remedy for piercing problems caused by fragile needles or for any application where penetration of a standard septum is an issue.
Chromacol K-Series Vial Closure
Description Quantity Part Number
Chromacol 9mm, Magnetic, Open Top Short Screw Cap with White Silicone/Red PTFE Septum 100/PK 9-MSC(BG)-ST1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red Rubber/Clear PTFE Septum 500/PK 9-SCK(B)-8RT1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red Rubber/Clear PTFE, Pre-slit Septum 500/PK 9-SCK(B)-8RT1X
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, White Silicone/Red PTFE Septum 500/PK 9-SCK(B)-ST1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, soft Blue Silicone/PTFE Septum 500/PK 9-SCK(B)-ST101
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, White Silicone/Red PTFE, Pre-slit Septum 500/PK 9-SCK(B)-ST1X
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red PTFE/White Silicone/Red PTFE Septum 500/PK 9-SCK(B)-TST1
Chromacol 9mm Solid Top Short Screw Cap (Blue), Polypropylene, Red Rubber/Beige PTFE Septum 500/PK 9-SCSK(B)-8RT1
Chromacol 9mm Short Screw Cap (Blue), Polypropylene with Integral Polypropylene Membrane 500/PK 9-SCNK(B)
Chromacol Convenience Kit, Wide Open Short Screw Vial, Clear, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK 2-SVW8-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK 2-SVWST-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Amber, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK 2-SVW(A)8-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Amber, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK 2-SVW(A)ST-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Agilent LC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK HPLSK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Thermo GC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, soft Blue Silicone/PTFE Septum 100/PK TTRK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Varian GC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK VAGK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Varian LC Autosampler, Clear, Patch, Blue 9 mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK VALK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Waters Alliance LC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK WALK
2Soft durometer septa are a effective remedy for piercing problems caused by fragile needles or for any application where penetration of a standard septum is an issue.
National Target DP 9mm C5000 Series Vial Closure
Description Quantity Part Number
National Target DP Blue 9mm Screw thread Cap with Natural PTFE/Blue Soft Silicone Septum2 100/PK C5000-44B
National Target DP Blue 9mm Screw thread Cap with Natural PTFE/Blue Soft Silicone Septum, Pre-slit2 100/PK C5000-45B
National Target DP Blue 9mm Screw Thread Cap with Ivory PTFE/Red Rubber Septum 100/PK C5000-51B
National Target DP Blue 9mm Screw Thread Cap with Red PTFE/White Silicone/Red PTFE Septum 100/PK C5000-53B
National Target DP Blue 9mm Screw Thread Cap with Red PTFE/White Silicone Septum 100/PK C5000-54B
National Target DP Blue 9mm Screw Thread Cap with Blue PTFE/White Silicone Septum, Preslit 100/PK C5000-55B
National Target DP Clear 9mm Screw Thread Cap with Integral Polypropylene Membrane 100/PK C5000-50
National Target DP Blue 9mm Screw Thread Cap with Integral Polypropylene Membrane 100/PK C5000-50B
National Target DP Blue 9mm Screw Thread Closed Top Cap with Ivory PTFE/Red Rubber Septum 100/PK C5000-99B
National Convenience Kit, Clear Glass, Blue DP Cap w/ Ivory PTFE/Red Rubber Septum 100/PK C5000-80
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Ivory PTFE/Red Rubber Septum 100/PK C5000-80W
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Red PTFE/White Silicone/Red PTFE Septum 100/PK C5000-86W
National Convenience Kit, Clear Glass, Blue DP Cap w/ Red PTFE/White Silicone Septum 100/PK C5000-92
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Red PTFE/White Silicone Septum 100/PK C5000-92W
National Convenience Kit, Clear Glass w/I-D Patch, Blue DP Cap w/ Blue PTFE/White Silicone pre-slit Septum 100/PK C5000-95W
National Assembled Vial Kit, Clear Glass, Ivory PTFE/Red Rubber Septum, Blue DP Cap 100/PK C5000-180
National Assembled Vial Kit, Clear Glass w/I-D Patch, Ivory PTFE/Red Rubber Septum, Blue DP Cap 100/PK C5000-180W
National Assembled Vial Kit, Clear Glass w/I-D Patch, Red PTFE/White Silicone/Red PTFE Septum, Blue DP Cap 100/PK C5000-186W
National Assembled Vial Kit, Clear Glass w/I-D Patch, Red PTFE/White Silicone Septum, Blue DP Cap 100/PK C5000-192W
2Soft durometer septa are a effective remedy for piercing problems caused by fragile needles or for any application where penetration of a standard septum is an issue.
Chromacol K-Series Vial Closure
Description Quantity Part Number
Chromacol 9mm, Magnetic, Open Top Short Screw Cap with White Silicone/Red PTFE Septum 100/PK 9-MSC(BG)-ST1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red Rubber/Clear PTFE Septum 500/PK 9-SCK(B)-8RT1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red Rubber/Clear PTFE, Pre-slit Septum 500/PK 9-SCK(B)-8RT1X
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, White Silicone/Red PTFE Septum 500/PK 9-SCK(B)-ST1
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, soft Blue Silicone/PTFE Septum 500/PK 9-SCK(B)-ST101
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, White Silicone/Red PTFE, Pre-slit Septum 500/PK 9-SCK(B)-ST1X
Chromacol 9mm Open Top Short Screw Cap (Blue), 6mm Opening, Polypropylene, Red PTFE/White Silicone/Red PTFE Septum 500/PK 9-SCK(B)-TST1
Chromacol 9mm Solid Top Short Screw Cap (Blue), Polypropylene, Red Rubber/Beige PTFE Septum 500/PK 9-SCSK(B)-8RT1
Chromacol 9mm Short Screw Cap (Blue), Polypropylene with Integral Polypropylene Membrane 500/PK 9-SCNK(B)
Chromacol Convenience Kit, Wide Open Short Screw Vial, Clear, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK 2-SVW8-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK 2-SVWST-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Amber, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK 2-SVW(A)8-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial, Amber, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK 2-SVW(A)ST-CPK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Agilent LC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, Red Rubber/ Clear PTFE Septum 100/PK HPLSK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Thermo GC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, soft Blue Silicone/PTFE Septum 100/PK TTRK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Varian GC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK VAGK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Varian LC Autosampler, Clear, Patch, Blue 9 mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK VALK
Chromacol Convenience Kit, Wide Open Short Screw Vial for Waters Alliance LC Autosampler, Clear, Patch, Blue 9mm Open Top Short Screw Cap, White Silicone/Red PTFE Septum 100/PK WALK
2Soft durometer septa are a effective remedy for piercing problems caused by fragile needles or for any application where penetration of a standard septum is an issue.
Resources