Support status tables
Key to the tables
Support Class Media Availability Future Service Packs Technical Support
Current Main Release and Current Service Pack Regularly Complete
Recent Main Release and Current Service Pack Usually Complete
Discontinued If stock available Rarely Limited
Obsolete None Never Limited
Thermo Scientific Dionex Chromeleon 7 Chromatography Data System
Release Most Recent
Service Pack
Support
Class
Tested Windows® OS Releases Tested Database Releases
7.2
July
2013
7.2 SR4
January
2016
7.2 SR3
September
2015
7.2 SR2 MUa
March
2015
7.2 SR2
December
2014
7.2 SR1 MUa
August
2014
Current Windows® 8 Pro (64 bit)
(SR3 and above)
Windows® 7 SP1(32 and 64 bit)
Professional Edition
Ultimate Edition
Windows Server® 2008R2
Windows Server® 2012R2
Windows® XP SP3 (32 bit only)
Professional Edition
(not tested for 7.2 SR1 or later)
SQL Server® 2005 Express
Version 9.3.4035.00
SQL Server® 2008 R2 Express SP2
Version 10.50.4000.0
SQL Server® 2008 R2
Standard Edition
Version 9.3.4035.00
Microsoft SQL Server 2012
Standard Edition
Oracle® 11g
Standard Edition
Version 11.2.0.1.0
7.1
January
2011
SR3
December
2013
SR2
October
2012
SR2 MUb
March
2013
Recent Windows® 7 (32 and 64 bit)
Professional Edition
Ultimate Edition
Windows Server® 2008R2
Windows® XP SP3 (32 bit only)
Professional Edition
SQL Server® 2005 Express
Version 9.3.4035.00
SQL Server® 2008 R2
Standard Edition
Version 9.3.4035.00
Oracle® 11g
Standard Edition
Version 11.2.0.1.0
7.0
June
2009
SR1
May
2010
Obsolete Windows Vista® SP1
Windows® XP SP3
SQL Server® 2005 Express
Version 9.2.3042.00
Thermo Scientific Dionex Chromeleon 6 Chromatography Data System
Release Most Recent
Service Pack
Support
Class
Tested Windows® OS Releases Tested Database Releases
6.80
June
2006
DU15a
May
2015
SR15
October
2014
SR14
August
2014
Current Windows® 7 (32 and 64 bit)
Professional Edition
Ultimate Edition
Windows Vista® (32-bit)
(Vista SP1 minimum)
(Vista SP2 with SR8 and above)
Business Edition
Ultimate Edition
Windows® XP SP3
(only SP3 and above)
Windows® 2000 SP4
(only SP3 through SP9)
Windows® 2003 Server
(only SP2 and above)
Windows® 2008 Server R2
(only SR10 and above)
Windows® 2008 Server R2 SP1
(only SR13 and above)
Oracle® 8.1.7
Oracle® 9.0.1.1.1
Oracle® 11.2.0.3.0
SQL Server® 2000
Version 8.00.382
SQL Server® 2005
Version 9.00.1399.00
SQL Server® 2008
Version 10.00.1600
SQL Server® 2008 R2
Version 10.50.1600.1
SQL Server® 2008 R2 SP1
Version 10.50.2500.0
6.70
April
2005
SP10
April
2009
Obsolete Windows® XP SP1, SP2
Windows® 2000 SP2, SP3, SP4
Windows Server® 2003 SP1
Oracle® 8.1.7
Oracle® 9.0.1.1.1
SQL Server® 2005
SQL Server® 2000
version 8.00.382
6.60
March
2004
SP9
December
2007
Obsolete Windows® XP SP1, SP2
Windows® 2000 SP2, SP3, SP4
Windows Server® 2003
Oracle® 8.1.7
Oracle® 9.0.1.1.1
SQL Server® 2000
version 8.00.382
6.50
January
2003
SP10
April
2006
Obsolete Windows® XP SP1
Windows® 2000 SP1, SP2, SP3
Windows® NT 4.0 SP6a
Oracle® 8.1.7
Oracle® 9.0.1.1.1
SQL Server® 2000
version 8.00.382
6.40
November
2001
SP8
December
2004
Obsolete Windows® 2000 SP1, SP2
Windows® NT 4.0 SP6a
Windows® ME
Windows® 98 Second Edition
Oracle® 8.1.7
SQL Server® 2000
Version 8.00.382
SQL Server® 7
6.30
June
2001
SP8
October
2003
Obsolete Windows® 2000 SP1, SP2
Windows® NT 4.0 SP6a
Windows® ME
Windows® 98 Second Edition
Oracle® 8.1.7
SQL Server® 2000
version 8.00.382
SQL Server® 7
6.20
December
2000
SP9
December
2004
Obsolete Windows® 2000
Windows® NT 4.0 SP4
Windows® ME
Windows® 98 Second Edition
Windows® 95
information not available
6.10
May
2000
SP2
September
2000
Obsolete Windows® 98 Second Edition
Windows® 95
information not available
6.01
January
2000
None Obsolete Windows® 98 Second Edition
Windows® 95
information not available
6.00
October
1999
None Obsolete Windows® 98 Second Edition
Windows® 95
information not available
<6.00 Earlier None Obsolete Windows® 98 or earlier information not available
Copyright notice:
Windows, Windows Server, Windows Vista, and SQL Server are either registered trademarks or trademarks of Microsoft Corporation in the United States and/or other countries.
Oracle and Java are registered trademarks of Oracle and/or its affiliates. Other names may be trademarks of their respective owners.