hdi-ase-350-accelerated-solvent-extractor-230x195
Get to the results faster while saving time, solvent and money
Using organic acids and aqueous solvents or acids and bases at high temperatures and pressures, the Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extraction system extracts compounds from solid and semi-solid samples quickly with small solvent volume. You will save time, solvent, and money, and generate results in a fraction of the time compared to traditional techniques such as Soxhlet or sonication.
Popular accelerated solvent extraction products
featured-ase-350-230x100
Common applications of accelerated solvent extraction
Environmental samples
  • Pesticides/herbicides
  • PAHs/semivolatiles
  • PCBs
  • Dioxins/furans
  • TPH (DRO)
  • Explosives
  • Air sampling cartridges
  • Brominated flame retardants
Pharmaceutical samples
  • Active ingredients
  • Drug delivery devices/packaging
Polymer samples
  • Oils/organic acids
  • Xylene solubles
  • Polymer additives
Food samples
  • Vitamins/antibiotics
  • Contaminants
  • Natural products
  • Pesticides/herbicides
  • Pesticide residues
  • Fats/lipids
  • Herbal/dietary supplements
Consumer products
  • Detergents
  • Paper and pulp
  • Biofuels
  • Textiles and fibers
Featured accelerated solvent extraction categories
The Dionex ASE system performs extraction in your choice of stainless steel or Thermo Scientific™ Dionium™ extraction cells. They are available in a variety of sizes to suit your extraction requirements.
Worn extraction cell parts have to be periodically replaced to ensure a proper seal and to maximize the efficiency of your Dionex ASE extraction system. Parts include extraction cell bodies, PEEK seals, snap rings, and much more.
Filters and thimbles hold solid and semisolid samples in the extraction cells, prevent plugging of the fluid path, maximize separation of solvent, and make extraction cell cleaning easier. Filters are available in high-quality cellulose for routine use and in glass fiber for specialized applications.
Sorbents and dispersants are used to improve recovery of analytes during or after extraction. These Dionex ASE-Prep products disperse the sample matrix, remove moisture, neutralize mineral acids and bases, or clean up extracts after extraction.
On Demand Webinars
On Demand Webinars
Accelerated Solvent Extraction Application Notebooks
Dionex ASE Accelerated Solvent Extraction application notebooks are now available for download:
  • Food Safety ASE Applications
  • Environmental ASE Applications
  • Environmental ASE Technical Resource Guide
Download Application Notebook ›
Resources
Application Notes
Literature
Videos