Gas Chromatography
Automate the GC workflow—from sample preparation to results
From sample preparation and advanced sampling handling functions, to modern GC systems, quality columns and state-of-the-art reporting, these solutions are used by food, environmental, pharmaceutical, industrial and other fields to improve manufacturing processes and protect people and the environment from harmful contaminants.
Popular gas chromatography products
Featured gas chromatography categories
These state-of-the-art systems feature instant-connect injectors and detectors. Use as standalone systems or couple with mass spectrometry.
Solutions for the most challenging applications. Meet stringent requirements for performance, reliability, and value with these powerful instruments.
This instant-connect injector offers maximum flexibility to meet all types of routine and high throughput analytical needs. You can switch instant-connect injectors in 2 min.
You can switch an instant-connect detector in 2 min. They include thermal conductivity, flame ionization, pulsed discharge, nitrogen phosphorus, and electron capture detectors.
These easy-to-use systems perform a range of sample handling functions spanning liquid, headspace, and SPME injections, as well as dilutions, additions, and derivatization functions.
Featuring high temperature stability, low bleed and long lifetimes, our proven GC columns range from polar to non-polar to specialty GC columns for a multitude of GC or GC-MS applications.
Here you will find gas filters, injection port accessories, GC-specific syringes, tools, tubing and GC sample handling products such as derivatization systems and vials and closures.
Offering high-purity silylation and acylation derivatization reagents and siliconizing flluids for various GC and GC-MS applications.
Featured chromatography video
Watch to learn about the benefits and features of the Thermo Scientific TRACE 1300 Series Gas Chromatograph.
Need to find GC columns or accessories?
Access the latest version of our Chromatography Columns and Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog