Engineered for superior performance
Use our inert, low bleed 100% dimethyl polysiloxane Thermo Scientific™ GC columns to ensure reliable, reproducible results for your GC and GC-MS analyses of nonpolar compounds in a wide variety of samples including  petrochemicals, environmental pollutants, pesticides, essential oils, drugs of abuse, among many others.
Non-polar gas chromatography columns
Common applications:
  • Chlorinated and nitroaromatic compounds
  • Hydrocarbons
  • Polychlorobiphenyls (PCBs)
  • Refinery gases
  • Gasoline range organics (GROs)
  • Drugs of abuse
  • Pesticides
  • Essential oils
Column
Recommended Application
TraceGOLD™ TG-1MS GC Columns Achieve consistent results with this low bleed, stable column which delivers optimal signal-to-noise ratio, sensitivity and MS integrity for a wide variety of applications.
TraceGOLD™ TG-1MS GC Columns with SafeGuard Extend column life with the integrated guard of the this stable, low bleed GC column recommended for the analyses of petrochemicals (including GROs or gasoline range organics), refinery gases, pesticides and essential oils.
TraceGOLD™ TG-1MT Metal GC Columns Push the limits of your analyses with these nonpolar, metal columns which allow you to work up to 430˚C with no compromise in performance when analyzing highly active and aggressive compounds.
TRACE™ TR-1MS GC Columns Use this low bleed non-polar column which is excellent for GC-MS environmental analyses that require an inert phase that can operate at temperatures as high as 340–360°C.
Need to find GC columns or accessories?
Access the latest version of our Chromatography Columns and Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog