syncronis-hplc-columns-showcase-image-230x195
Enter a new world of chromatography
When developing a new method, one of the most important goals for the chromatographer is to achieve a consistent, reproducible separation. This makes selecting an HPLC column that offers highly-reproducible results essential to attaining this goal. 
Features of Syncronis columns
Thermo Scientific Syncronis LC columns achieve high levels of consistency and reproducibility through:
  • Highly-pure, high-surface-area silica
  • Dense bonding and double endcapping
  • Stringent manufacturing and testing protocols
Syncronis column phases
Reversed phase
Normal phase
HILIC phase
Resources
Application notes
Catalogs & technical guides