Proven systems that are rugged and versatile, with the flexibility to customize
Meet the needs and requirements for your application by selecting from our extensive portfolio of Thermo Scientific UltiMate 3000 HPLC systems. With customizable configurations, these systems provide you with the flexibility to accomplish all applications from method development to routine analysis with HPLC to UHPLC capabilities.
Discover which UltiMate 3000 system is right for you
The UltiMate 3000 HPLC series provides excellent chromatographic performance while maintaining easy-to-use, reliable operation. Compatibility with any of our columns offers maximum application flexibility to meet all your separation needs. Regardless of the configuration you choose you will receive a highly integrated setup with optimized fluidic connections, as well as the industry leading easy-to-use Thermo Scientific Chromeleon Chromatography Data System (CDS) software. Our Chromeleon CDS is the preferred software for single-point intelligent control, unique data processing tools, and intelligent run automation functionality. In addition, we offer integration solutions for other CDS packages.
UltiMate 3000 HPLC and UHPLC systems comparison
    Rapid Separation (RS) Standard
(SD)
Basic Semi-
preparative
Performance
System pressure
Retention time precision
Flow range
Low dispersion and high throughput
Features
Design Modular
flexible
Modular
flexible
Modular
flexible
Modular
flexible
Pump options Binary
Quaternary
Dual
Binary
Quaternary
Dual
Quaternary
Isocratic
Binary
Biocompatible options  
Active heating column compartment
Sample cooling        
Column capacity
Viper virtually zero dead volume system connections      
Adjustable gradient delay volume (depending on mixer configuration)     Quaternary Basic:    
Isocratic Basic: n.a.
Application Range
Fraction collection*        
Tandem LC*    
Application switching*    
Inverse gradient for uniform response*    
Automated on-line SPE*    
Automated method development*    
Large volume injection
*Requires additional equipment Thermo Scientific Viper UHPLC Fittings Kits
Order all the UHPLC fittings and capillaries you need for your system or application in a single kit
= Not applicable = Poor = Good = Better = Best
Compatible components for all UltiMate 3000 HPLC and UHPLC systems
Multiple Detector Options HPLC and UHPLC Columns Chromeleon CDS Software
  • Charged aerosol detector
  • Diode array detector
  • Electrochemical detector
  • Fluorescence detector
  • Mass spectrometry detection
  • Multiple wavelength detector
  • Refractive index detector
  • Variable wavelength detector
  • An extensive portfolio of columns
  • In a broad variety of inner diameters, lengths, particle sizes, and chemistries
  • Streamlines your data processing
  • Industry-leading ease of use and intelligent functionality
  • Unique user guidance and hardware automation features designed for UltiMate 3000 systems
Not sure which system is right for you?
Connect with an expert to help you decide
Contact us
Monitor your system remotely from anywhere in the world
  • Connect your lab to boost productivity with secure remote access to your data and instruments
  • Real-time monitoring of your instrument with automatic email notifications
  • Store data in secure accounts and share with colleagues and collaborators
Learn more ›