iQ Series Gas Analyzers for Ambient Air Monitoring
The environment is a continuous source of critical information. And with the new iQ Series of Gas Analyzer, so is your instrument.
Proactive communications and predictive diagnostics help eliminate unexpected trips to the field, so you can plan maintenance tasks around your schedule. The iQ Series has a touch screen display that's easy to operate, and it's engineered to get smarter over time.
Request a quote
Predictive diagnostics
Address problems before they occur with improved serviceability and plug and play components. 24/7 access to your analyzer’s information helps minimize troubleshooting time with innovative, interchangeable DMC modules in a highly precise, yet tough solution.
Proactive communications
Maintain optimum performance through iQ360 and email alerts. When subscribed to iQ360 service, your instrument communicates directly to our technical support team upon alarm activation. Our team develops a corrective plan and emails it to your staff. Eliminate surprises and take control of your day with the iQ Series Gas Analyzers.
Personal device connectivity
Real time, continuous remote monitoring is now possible with the iQ companion mobile device app. The iQ app allows for remote monitoring of iQ gas analyzers, simplified ways of getting in touch with us, and instant access to both maintenance alerts and product documentation.  
iQ Series Gas Analyzers App
Download the app
Featured gas analyzers for ambient air monitoring
The Thermo Scientific™ 43iQ Sulfur Dioxide Analyzer provides the same level of performance when measuring sulfur dioxide (SO2) in ambient air up to 100ppm using pulsed fluorescence technology.
The Thermo Scientific™ 48iQ Carbon Monoxide Analyzer provides the same level of performance when measuring the amount of carbon monoxide (CO) in ambient air using the gas filter correlation technique.
The Thermo Scientific™ 49iQ Ozone Analyzer provides the same level of performance when measuring the amount of ozone in ambient air using dual-cell, UV photometric technology.
The Thermo Scientific™ 49iQ Primary Standard Ozone Analyzer provides the same level of performance when producing ozone concentrations of up to 5000ppb using dual-cell, UV photometric technology.
The Thermo Scientific™ 111iQ Zero Air Supply supplies pollutant-free air (zero air) from ambient air to allow for proper zeroing, and to provide clean diluent air for spanning ambient air analyzers.
Where some see data, we see intelligence
Resources
Product support and services