air-quality-showcase-image-239x195
Making the air, cleaner, safer, healthier
Ensuring clean air requires collaborative effort in the United States, China, India, Europe and all points of the globe. Governmental agencies are working harder than ever toward creating regulations that minimize the release of pollutants and harmful toxins in the air. For manufacturers and industrial plants, successfully complying with those regulations requires equipment with advanced technology and maximum reliability.
Contact an application specialist
Subtopics
TOEM Technology for Particulate Matter Measurement
Particulate matter monitoring systems that employ TEOM technology are “gravimetric” instruments that draw ambient air through a filter, weighing the filter and calculating concentrations of particulate matter. 
Beta Attenuation Technology for Particulate Matter Measurement
The principle behind beta attenuation particulate sampling instruments is that energy is absorbed from beta particles as they pass through particulate matter collected on a filter media. 
Global Air Quality Regulations
cement-coal-minerals-125x125
Cleaner air for China
As China experiences dangerously high levels of particulate matter and other toxins in the air, negative health effects on residents are a top concern.  Ozone, carbon dioxide, nitrogen oxides, and particulate dust particles are harmful substances when inhaled.  Particulate matter that is very small, smaller than 2.5 microns in diameter, is referred to as PM2.5 and it can enter deep into the lungs causing cancer and other lung disease.  Measuring the pollutants in the air is a critical step toward cleaner air.  The most reliable and accurate technology for measuring PM 2.5 are tapered element oscillating microbalances (TEOM) and beta attenuation technologies.
Downloadable Infographic—Adhering to the US EPA MACT rule
The U.S. EPA requires compliance as set forth under the Maximum Achievable Control Technology(MACT)/National Emission Standards for Hazardous Air Pollutants Compliance Monitoring (NESHAP) rule for Portland cement manufacturing. If you're a facility that must adhere to this U.S. EPA clean air standard, download our MACT compliance infographic.
Fast5 Tips and Tricks for Gas Analyzer and CEMS
If you have an onsite challenge, chances are you will find the solution in one of our Fast5 issues. Sign up here to read previous issues or to receive future issues. Topics include preparing for ozone season, extending calibrator certification, and optimizing the measurement of NO, NO2 and NOx.
Talk Tech
Find solutions and tips to ambient particulate challenges in Talk Tech Q&A. Topics include cold weather monitoring and instrument maintenance recommendations.
Complying With Mercury Emissions Regulations
In order to comply with Under the Interim EPA Traceability Protocol, there are four approved methods to quality check a mercury generator: field reference generator, permeation source, sorbent tube and mercury gas cylinder. While each method provides the same level of compliance, the permeation tube method is the best way to extend the certification. The permeation tube generates a known and reliable concentration of mercury at a constant temperature and flow. This allows for automated audits confirming the reliability of the calibrator output.
Environmental Resource Library
Access a targeted collection of scientific application notes, case studies, videos, webinars and white papers for air quality, contaminant and water quality analysis.
External resources
Support
See all Learning Centers ›
Share
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.