Fast5-banner-740x142
July 2014
Volume I, Edition 7
Mercury continuous emissions monitoring systems (CEMS) are primarily designed for emissions compliance monitoring, but many facilities find the systems valuable in monitoring process parameters. Here are some key findings:
C-Series analyzers
Sales and support for the Thermo Scientific C-series Gas Analyzers will be discontinued beginning August 1, 2015. 
Read discontinuation notice
Data is everything
fast5-one
Obtaining meaningful data can have a significant impact on your process whether using the mercury CEMS for process monitoring or compliance monitoring. Accurate and relevant data can result in cost and resource savings. The table below lists some of the required tests and recommended on- going checks using the Mercury CEMS for either compliance or process control in order to provide the greatest amount of meaningful data.
Note that the acceptance criteria for compliance monitoring falls under the MATS or the PC MACT rule and this is not a complete list. For process control applications, the pass/fail criteria can be user-specific depending on the application.
System Check / Configuration
Compliance Monitoring
Process Control
Zero & Span
Required Daily
Strongly Recommended Daily
System Integrity Test
Required Weekly
Optional
Linearity
Required Quarterly
Recommended
Elemental Generator Audit
Required Monthly
Recommended Semi-Annually
RATA
Required Annual
Partial RATA Recommended
Speciation Mode
Total Hg Only Required
Speciation Recommended
7 Day Drift Test
Required for Initial Certification
Recommended upon install
Cycle response time
< 15 minutes
Required for Initial Certification
Recommended upon install
Probe Type
Multiple Options
Inertial Probe Recommended for dirty applications
Tightening control over your process
fast5-two
Having a real-time monitor running in your process enables full visibility that can help tighten your process parameters. Any Mercury control performance guarantee supplied by a sorbent manufacturer can be verified when the probe is upstream of the mercury control system. When the probe is downstream of the mercury control system, the amount of sorbent can be easily and accurately adjusted to reach your compliance goals. The Mercury Freedom System can be a good solution for compliance monitoring in addition to process monitoring because it is a self-contained system and no other hardware or software is needed.
Save on consumable costs
fast5-three
Facilities using mercury CEMS for process control realize significant savings in consumable costs. Whether using sorbent injection, activated carbon injection or other plant-specific techniques for control of mercury, having a real-time monitor integrated into the process eliminates the guess work associated with sorbent injection. Using a real-time monitor reduces the risk of under-injecting and minimizes the risk of over-injecting sorbent. As the mercury level gets more accurate, the realization of cost savings increases. 
Put the personnel behind your process
fast5-four
Having a technician who is trained in mercury CEMS is one avenue toward consistent and successful mercury monitoring. It has been shown that facilities with an onsite mercury CEMS expert can diagnose performance issues more quickly and accurately. Furthermore, properly trained mercury technicians are more likely to bring the issue to resolution quicker. 
Technical training for all
fast5-five
All technicians who interact with the mercury CEMS benefit from training. There is still room available in the August and November mercury CEMs course  in Franklin, MA. These courses are designed for users of varying abilities and cover regulatory considerations, installation and configuration, sample probe use, and advanced maintenance and troubleshooting. Customized classes are available and easily scheduled by contacting our Technical Training team.
fast5-arrow
Next Month's Topic: Tips for measuring nitrogen oxides (NOx) levels in the air
fast5-showcase-image-230x195
Join Fast5 today
Our technical and engineering staff share their product and application expertise for gas analyzers and systems. 
Sign up today to receive Fast5 tips and tricks every month! 
Sign up ›
Resources
Support
Factory repair
Schedule your analyzer, calibrator, probe controller or probe for a Return Material Authorization (RMA) to factory depot repair for complete maintenance and factory specification service.
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.