gino-tellier
Gino Tellier
Technical support specialist
Environmental and process monitoring
Meet our techs: Gino Tellier
Gino brings over 30 years of expertise in service, quality assurance, manufacturing, operations and technical support to our customers looking to optimize in ambient particulate applications.
Our techs are here to answer your questions, just email us.
Ask our techs ›
If you like this month's Q&A, check out our Talk Tech archive.
Question
 If you have a Thermo Scientific 1405 TEOM™, Continuous Ambient Particulate Monitor or 1405-DF TEOM™ Continuous Dichotomous Ambient Air Monitor, it is recommended to service the switching valve every year. Sometimes you need to service the switching valve ahead of schedule if you find a leak in the system caused by the “V” seals being damaged prematurely or if you discover that the motor is not moving the inner block between its 2 positions during the switching process.
The switching valve maintenance is best performed annually at the time of the dryer replacement and cooler cleaning process to minimize downtime and to maximize the performance of a complete maintenance schedule.
See Technical Bulletin for switching valve service instructions on the new valve design.
See Technical Bulletin for switching valve service instructions on the original valve design.
tech-talk-showcase-image-230x195
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.
Subscribe to Talk Tech ›
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.