talk-tech-sept-showcase-image-230x195
Debbie Bowe
Sr. Tech Specialist and Master Trainer
Environmental and Process Monitoring
Meet our techs: Debbie Bowe
Debbie enjoys helping customers solve problems in all areas including programming samplers, passing leak checks and getting flow audits to meet standards.
Our techs are here to answer your questions, just email us.
Ask our techs ›
If you like this month's Q&A, check out our Talk Tech archive.
Question
Answer
Recent changes to Microsoft Windows Security interfere with the installer program. You must disable 'Windows firewall' and if you have a wireless network adapter disable the wireless adapter temporarily.  If your current firmware version is 1.56 or higher you can update to the current release, which is version 1.71 as of March 2015. 
If your current firmware version is 1.55 or earlier, you must update to 1.56 first before updating to the current release.
Read through the read me file that accompanies the firmware files for upload instructions and reference to the new Program Register Codes for use with current data acquisition systems. The unit must also be set to 'EPA' designation in the advance settings for mass calculation variables. PM10 concentrations are calcuated for all reporting conditions, Standard , Volumetric and FEM and listed by variable or register.
For additional compatibility issues, refer to the Windows® 7 and Windows® 8 Software Compatibility Technical Bulletin.
tech-talk-showcase-image-230x195
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.
Subscribe to Talk Tech ›
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.