Perfect your food pathogen testing workflow
The Pathatrix® instrument is a fully automated sample prep system designed to rapidly and accurately handle any sample matrices to help save you time and money. Using paramagnetic particles that are either antibody-coated or positively charged, the Pathatrix® system selectively binds and concentrates target organisms from many of the complex matrices found in food, environmental, and biothreat samples.
Request more info or a demo
Watch our free on-demand webinar | Request a Pathatrix® demo in your lab | Pathatrix® brochure
The Pathatrix® Auto System is fully compatible with a wide range of detection technologies. The captured, concentrated, and cleaned sample produced by the Pathatrix® system helps ensure a more reliable and accurate result from your chosen detection method. The time to result is also reduced due to the concentration effect of the Pathatrix® system. Simple linking protocols are available for all of the commonly used detection technologies, including:
  • Agar plates—selective and chromogenic
  • Colorimetric assays—ELISA and lateral flow
  • PCR—real-time or conventional
Key features and benefits
Cleans and concentrates target—helps reduce false positives and indeterminate results
Pooling of up to 10 samples—save up to 90% on downstream detection costs
15-minute, fully automated run-time—quicker time to results and improved productivity