In Food Safety and Food Testing, Never Sacrifice Product Quality for a Fast Release.
Increase your product quality and decrease your time to market with Life Technologies Salmonella testing solutions.
  • Release your product fast and minimize your inventory holdings - accurate and specific detection of Salmonella in as little as 18 hours.
  • Lower chance of false positives  - highly specific molecular TaqMan® PCR assays limit cross reaction with near neighbor organisms.
  • Confidence in your results - external approvals and validation: MicroSEQ® Salmonella spp AOAC & AFNOR validation, TaqMan® Salmonella Enteritidis FDA Equivalent & NPIP approved.
Step-by-Step Guide to Salmonella Testing Solutions
Universal spin-column-based format for use with all food sample types. Bead-based chemistry for the high-quality purification of DNA from a variety of food samples. Magnetic particle processing instrument for the rapid processing of up to 96 samples.
Recommended Products
PrepSEQ® Rapid Spin Sample Preparation Kits
PrepSEQ® Nucleic Acid Extraction Kit
MagMax™ Express 96 Instrument
Complete, validated assays into a single reaction bead. Simply add your purified DNA and get your results in this easy-to-use format. Premier qPCR instrumentation from a leader in molecular analysis equipment. Utilize fast qPCR chemistries to process 96 samples typically in under an hour.
Recommended Products
MicroSEQ® Salmonella spp.detection Kit
Taqman®Salmonella enteritidis detection Kit
Applied Biosystems 7500 Fast Real-Time PCR Instrument
Taqman® Salmonella enterica & Prepman® Ultra
RapidFinder™ Express Software works with your 7500 Fast Real-Time PCR Instrument to provide automated pathogen detection calls and report generation. Minimizes the chances for user error or operator bias.
Recommended Products
7500 Fast Real-Time PCR System
RapidFinder™ Express Software
Offical methods, such as those from the FDA BAM or ISO, are recommended for culture confirmation of results.
Taqman® Salmonella Enterica detection Kit has been determined to be equivalent to the FDA BAM method in accuracy, precision and sensitivity for testing egg and poultry environmental samples.