AmpFℓSTR® Identifiler® Plus PCR Amplification Kit
Optimized for casework, the AmpFℓSTR® Identifiler® Plus PCR Amplification Kit enables forensic analysts to recover more interpretable results from a wider range of DNA evidence samples with increased confidence.
The new kit includes the same primers and allelic ladder as the widely used Identifiler® Kit and harnesses next-generation PCR amplification technology to help provide enhanced sensitivity, a cleaner baseline, improved performance on mixtures, and the ability to overcome high levels of PCR inhibition.
Order now
Key Features
  • The ability to overcome high levels of PCR inhibition
  • Enhanced sensitivity
  • Improved performance on mixtures
  • Cleaner baselines with reduced artifacts
  • Works in conjunction with the Quantifiler® HP and Trio DNA Quantification Kits, and the MiniFiler™ PCR Amplification Kit to help maximize recovery from heavily degraded samples
  • Genotyping concordance with samples typed with the Identifiler®, Identifiler® Direct, Profiler Plus®, COfiler®, SGM Plus®, NGM™, NGM SElect™, NGM SElect™ Express, and recently introduced GlobalFiler™ and GlobalFiler™ Express amplification kits.
Resources
Related applications