Efficient DNA Collection, Storage, and Downstream Analysis
Human identification and forensic investigations depend on the careful collection and handling of samples. It is critically important that samples are collected using devices designed to:
  • Enable efficient, high-quality PCR amplification for genotyping
  • Facilitate automation, preservation, and storage
  • Maintain chain of custody and sample integrity
To meet this need, we offer innovative, high-quality collection solutions through a partnership with Copan Flock Technologies, a leader in manufacturing sample collection and DNA preservation tools. The Copan NUCLEIC-CARD™ and 4N6FLOQSwabs™ collection systems, in combination with our completely integrated human identification analysis workflows, enable forensic and paternity laboratories to maximize efficiency and achieve the highest quality results.
Featured
The NUCLEIC-CARD™ Sample Collection Devices consists of a plastic holder carrying a framed NUCLEIC-CARD™ COLOR (1 spot). The NUCLEIC-CARD™ COLOR matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature.
Learn more
ApplicationProducts
Reference Sample Collection NUCLEIC-CARD™ DNA Collection System
4N6FLOQSwabs™ DNA Collection System
Evidence Sample Collection 4N6FLOQSwabs™ DNA Collection System, Crime Scene