EPM_Level_Application_Shot-showcase-image-230x195
Demanding applications require proven technology
Accurate process material level measurement is necessary to maximize efficiency of industrial processes. For the most challenging applications, rely on non-contacting, easy to use, continuous process level gauges with a nuclear source. Nuclear gauges offer precise process material level measurements with proven stability at the widest temperature range and are designed to meet shock and vibration standards. These gauges are mounted outside of the vessel and the gamma energy emitted by the source is transmitted through the vessel walls. The electronics within the LevelPRO gauge converts the energy reading to an accurate and precise level measurement.
Contact an application specialist
Popular level measurement products
1400S Stainless Steel Transmitter for Signal Processing
4790 Continuous Gamma Level System
LevelPRO Series of Gauges
Featured level measurement products
Easy-to-use and offering accurate continuous level measurements using a smaller source, the Thermo Scientific™ LevelPRO Series of gauges are well suited for a variety of industrial applications.
Optimize delayed coking operations with the Thermo Scientific™ KRILPRO Neutron Backscatter Foam Level/Interface Device. Rugged and compact, this non-contacting device enables refiners to maximize coke drum output per cycle while significantly reducing anti-foam usage.
The Thermo Scientific™ Nitus Gamma Backscatter Gauge is a non-contacting gauge that provides highly precise online measurement of density and level in the largest tanks and pipes. It responds instantly to significant density and level changes to avoid tank overspills and other upset conditions.
PNF Point Level Switch
Mount the Thermo Scientific™ PNF Point Level Switch on tanks, bins, hoppers, pipes, chutes or other vessels to sense the level of liquids, slurries, and solids. It is is a non-contacting, non-invasive point level switch for high- or low-level indication.
Featured blog article
A Hybrid Approach to the Delayed Coking Process
Hydrocarbon processing in today’s petrochemical industry demands greater process control, as well as an increase in process efficiency and safety. Coking is a refinery unit operation that produces petroleum coke. When neutron backscatter and gamma transmission methods are used in conjunction, called “Hybrid Coking Level,” they can offer superior insight into the process changes than if either method was used alone.
Featured level measurement videos
Coker Control Hybrid Solution
The Thermo Scientific™ KRILPRO Neutron Backscatter Foam Level/Interface Device helps you effectively monitor the coking process and improve your profitability.
Resources