heat-exchangers-showcase-image-230x195
Efficient, economical cooling for your applications
Discover an efficient and economical alternative to traditional compressor-based chillers with Thermo Scientific™ heat exchangers. Benefit from a clean, stable, controlled, closed-loop water cooling system that rejects the process heat into an existing in-house facility water supply.
Our heat exchangers are designed to help eliminate problems associated with direct use of in-house water, such as insufficient or fluctuating flow, changing pressure, poor water quality, and temperature instability.
Take advantage of an existing in-house water system for heat removal. Thermo Scientific System Water-to-Water Heat Exchangers use less energy and cost less to operate than traditional compressor-based chillers. They provide a clean, stable, controlled, closed-loop water cooling system that rejects the process heat into an existing in-house facility water supply.
This helps eliminate the problems associated with the direct use of in-house water, such as insufficient or fluctuating flow, changing pressure, poor water quality, and temperature instability.
semi-processes-showcase-image-230x195
Supporting your semiconductor processes
Compact, reliable heat exchangers provide a low energy, efficient way to remove heat from water-cooled applications such as semiconductor testing.
Learn about our comprehensive portfolio of temperature control products
Access resource library
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.